Skip to Content
Merck

Skip To

1.07192

Pepsin

(from porcine gastric mucosa) cryst. lyophilized 10 FIP-U/mg for biochemistry EC 3.4.23.1

Synonym(s):

Pepsin

Sign In to View Organizational & Contract Pricing.

About This Item

CAS Number:
Molecular Weight:
35000
UNSPSC Code:
12352204
EC Index Number:
232-629-3
NACRES:
NA.21
MDL number:
eCl@ss:
42010127
Biological source:
Porcine gastric mucosa


biological source

Porcine gastric mucosa

Quality Segment

pH

4.0-5.0 (20 °C, 10 g/L in H2O)

solubility

10 g/L

bulk density

380‑420 kg/m3

storage temp.

2-30°C

General description

(from porcine gastric mucosa) cryst. lyophilized 10 FIP-U/mg for biochemistry EC 3.4.23.1
Protease
Pepsin is a member of the small family of closely related aspartic proteinase enzymes. Pepsin hydrolyzes proteins and petides, including those with bonds adjacent to aromatic or dicarboxylic L-amino acid residues; preferential cleavage: Phe- and Leu-.

Application

Pepsin (from porcine gastric mucosa) cryst. lyophilized 10 FIP-U/mg for biochemistry EC 3.4.23.1. CAS 9001-75-6, pH 4.0 - 5.0 (10 g/l, H₂O, 20 °C), molar mass 35000 g/mol.

Analysis Note

Appearance (description)fine-crystalline powderAppearance (color)almost whiteActivity (Hemoglobin; pH 1.6; 25 °C)≥ 10 FIP-U/mgpH-value (1 %; water)3.5 - 4.5Water (according to Karl Fischer)≤ 8 %Microbiological testpasses testColony count (aerobic bacteria)≤ 10000 CFU/gColony count (Yeasts and moulds)≤ 500 CFU/gSalmonella speciesabsent in 1 gE.coliabsent in 1 g

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
516360P6887SRE0001
biological source

Porcine gastric mucosa

biological source

-

biological source

Porcine gastric mucosa

biological source

-

storage temp.

2-30°C

storage temp.

−20°C

storage temp.

−20°C

storage temp.

−20°C

solubility

10 g/L

solubility

10 mM HCl: 0.5 mg/mL

solubility

deionized water: soluble 10 mg/mL, 10 mM HCl: soluble 4.0 mg/mL (Cold)

solubility

-

pH

4.0-5.0 (20 °C, 10 g/L in H2O)

pH

-

pH

-

pH

-

Quality Level

100

Quality Level

100

Quality Level

300

Quality Level

400

bulk density

380‑420 kg/m3

bulk density

-

bulk density

-

bulk density

-


Still not finding the right product?

Explore all of our products under Pepsin


signalword

Danger

Hazard Classifications

Eye Irrit. 2 - Resp. Sens. 1 - Skin Irrit. 2 - STOT SE 3

target_organs

Respiratory system

Storage Class

11 - Combustible Solids

wgk

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable

pictograms

Health hazardExclamation mark


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Global Trade Item Number

SKUGTIN
107192000104022536087856

Questions

Reviews

No rating value

Active Filters