Skip to Content
Merck

Skip To

1.06596

Sodium periodate

extra pure

Synonym(s):

Sodium (meta)periodate

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice
1 kg
Please contact Customer Service for Availability
CZK 9,030.00
25 kg

Estimated to ship onMay 25, 2026fromAreál Kühne+Nagel spol. s r.o.

CZK 180,400.00

About This Item

Linear Formula:
NaIO4
CAS Number:
Molecular Weight:
213.89
UNSPSC Code:
12352302
NACRES:
NA.21
MDL number:

CZK 9,030.00


Please contact Customer Service for Availability
Packaging: HDPE bottle

Request a Bulk Order
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Quality Segment

pH

5.2 (20 °C, 50 g/L in H2O)

mp

300 °C (dec.) (lit.)

SMILES string

[Na+].[O-]I(=O)(=O)=O

InChI

1S/HIO4.Na/c2-1(3,4)5;/h(H,2,3,4,5);/q;+1/p-1

InChI key

JQWHASGSAFIOCM-UHFFFAOYSA-M


Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
769517S18781.06597
Quality Level

200

Quality Level

100

Quality Level

300

Quality Level

300

pH

5.2 (20 °C, 50 g/L in H2O)

pH

-

pH

3.5-5.5 (25 °C, 107 g/L)

pH

5.2 (20 °C, 50 g/L in H2O)

mp

300 °C (dec.) (lit.)

mp

-

mp

-

mp

270 °C (decomposes)


Still not finding the right product?

Explore all of our products under Sodium periodate


signalword

Danger

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 1 - Eye Dam. 1 - Ox. Sol. 1 - Skin Corr. 1C - STOT RE 1

target_organs

thymus gland

Storage Class

5.1A - Strongly oxidizing hazardous materials

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable



Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

Reviews

No rating value

Active Filters