Skip to Content
Merck

Skip To

1.06129

MOPS

solid, buffer substance MPS

Synonym(s):

3-Morpholinopropane sulfonic acid, 3-(N-Morpholino)propanesulfonic acid, 4-Morpholinepropanesulfonic acid, MOPS

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Pack SizeSKUAvailabilityPrice
25 g
Please contact Customer Service for Availability
CZK 1,350.00
250 g
Please contact Customer Service for Availability
CZK 4,060.00
10 kg
Please contact Customer Service for Availability
CZK 99,400.00

About This Item

CAS Number:
Molecular Weight:
209.26
UNSPSC Code:
12161703
EC Index Number:
214-478-5
NACRES:
NA.21
MDL number:

CZK 1,350.00


Please contact Customer Service for Availability

Request a Bulk Order
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Product Name

3-Morpholinopropane sulfonic acid, buffer substance MPS

Quality Level

form

solid

potency

>2000 mg/kg LD50, oral (Rat)

pH

3-5 (20 °C, 111 g/L in H2O)

mp

277-281 °C (decomposition)

solubility

1440 g/L

bulk density

350 kg/m3

storage temp.

15-25°C

SMILES string

[S](=O)(=O)([O-])CCC[N+H]1CCOCC1

InChI

1S/C7H15NO4S/c9-13(10,11)7-1-2-8-3-5-12-6-4-8/h1-7H2,(H,9,10,11)

InChI key

DVLFYONBTKHTER-UHFFFAOYSA-N

General description

buffer substance MPS

Application

3-Morpholinopropane sulfonic acid buffer substance MPS. CAS 1132-61-2, pH 3 - 5 (111 g/l, H₂O, 20 °C).

Analysis Note

Assay (alkalimetric)≥ 99.0 %Identity (IR-spectrum)passes testUV-Absorption (250 nm, 50 g/l, 1 cm, water)≤ 0.05Chloride (Cl)≤ 0.005 %Sulfate (SO₄)≤ 0.005 %Heavy metals (as Pb)≤ 0.0005 %Na (Sodium)≤ 0.01 %Loss on Drying (105 °C)≤ 0.2 %Biological buffer, pKa at 20 °C7.20

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
47589869947M1254
Quality Level

100

Quality Level

100

Quality Level

200

Quality Level

200

form

solid

form

solid

form

powder or crystals

form

crystalline powder

storage temp.

15-25°C

storage temp.

15-25°C

storage temp.

-

storage temp.

room temp

solubility

1440 g/L

solubility

water: 1 M

solubility

H2O: 1 M at 20 °C, clear, colorless

solubility

water: 0.5 g/mL, clear, colorless

pH

3-5 (20 °C, 111 g/L in H2O)

pH

-

pH

2.5-4.0 (25 °C, 1 M in H2O)

pH

2.5-4 (25 °C, 209 g/L)

mp

277-281 °C (decomposition)

mp

-

mp

-

mp

-


Still not finding the right product?

Explore all of our products under 3-Morpholinopropane sulfonic acid


Storage Class

13 - Non Combustible Solids

wgk

WGK 1

flash_point_f

230.0 °F - closed cup

flash_point_c

110 °C - closed cup



Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library





Global Trade Item Number

SKUGTIN
106129025004022536073132
106129002504022536073125
106129901004022536692777

Questions

Reviews

No rating value

Active Filters