Skip to Content
Merck

Skip To

1.05397

Malt extract broth

GranuCult® prime, according to IFU, granular, for yeasts, for molds, pack of 500 g

Synonym(s):

Yeasts and molds medium

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Pack SizeSKUAvailabilityPrice
500 g
Please contact Customer Service for Availability
CZK 4,730.00

About This Item

UNSPSC Code:
41106213
NACRES:
NA.74
EC Index Number:
232-310-9
Manufacturer/tradename:
GranuCult® prime
Agency:
according to IFU
Ph:
4.8 (25 °C, 17 g/L in H2O, after autoclaving)
Form:
granular
Suitability:
molds, yeasts
Application(s):
food and beverages
microbiology

CZK 4,730.00


Please contact Customer Service for Availability

Request more information
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


biological source

plant

Quality Segment

agency

according to IFU

sterility

non-sterile

form

granular

packaging

pack of 500 g

manufacturer/tradename

GranuCult® prime

storage condition

dry media (Keep tightly closed.)

technique(s)

microbiological culture: suitable

color

yellow-brown

pH

4.8 (25 °C, 17 g/L in H2O, after autoclaving)

solubility

17 g/L

bulk density

590 kg/m3

application(s)

food and beverages
microbiology

storage temp.

15-25°C

suitability

molds, yeasts

General description

GranuCult® plus Malt extract broth contains polysaccharides which are used as an energy source and make the medium acidic. Mycological peptone serves as a nitrogen source. Lactic acid, tartaric acid, or antibiotics suppress bacterial growth and make the medium more selective. The pH value of the culture medium should be adjusted to 3.5 to obtain fungal counts and inhibit the growth of the bacterial flora. The malt extract broth can be modified for the selective cultivation of Aspergillus flavus. Furthermore, adding certain indicator dyes to malt extract agar allows the differentiation of yeast and bacterial colonies.

Application

Malt extract broth is suitable for the detection, isolation, and enumeration of fungi, particularly yeasts and molds, in various materials and for the cultivation of test strains for microbiological vitamin assays.

Features and Benefits

  • GranuCult® offers superior granulated culture media
  • Safe and sustainable due to reduced risks associated with fine dust and toxic substance inhalation, resulting in a safer work environment
  • Excellent wettability, solubility, and free-flowing properties
  • Convenient, with minimal component separation and clumping, even under warm or humid conditions
  • High batch-to-batch reproducibility
  • Prolonged shelf life of up to five years
  • High number of test strains exceeding all regulatory demands
  • Granulation technology allows many supplements to be included, with no need to add these separately

Analysis Note

Appearance (clearness): clear
Appearance (color): yellowish brown
pH-value (25 °C): 4.6 - 5.0
Growth promotion test in accordance with the current version of DIN EN ISO 11133.
Growth (Candida albicans ATCC 10231 (WDCM 00054)): good to very good
Growth (Saccharomyces cerevisiae ATCC 9763 (WDCM 00058)): good to very good
Growth (Saccharomyces cerevisiae ATCC 9080): good to very good
Growth (Geotrichum candidum DSM 1240): good to very good
Growth (Rhodotorula mucilaginosa DSM 70403): good to very good
Growth (Penicillium commune ATCC 10428): good to very good
Growth (Aspergillus brasiliensis (formerly A. niger) ATCC 16404 (WDCM 00053)): good to very good
Growth (Trichophyton ajelloi ATCC 28454): good to very good
Incubation: 7 days at 26 ± 2 °C, aerobic

Legal Information

GRANUCULT is a registered trademark of Merck KGaA, Darmstadt, Germany
NutriSelect is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

We offer two media types: the superior granulated GranuCult® and the cost-efficient powdered NutriSelect® culture media, depending on your needs. The designations basic, plus, or prime are added to indicate the quality control level, from basic quality control to standard QC plus to prime for full regulatory compliance.

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
M64091.053911.05391
form

granular

form

powder

form

powder

form

powder

application(s)

food and beverages, microbiology

application(s)

bioburden testing
food and beverages, microbiology

application(s)

microbiology

application(s)

microbiology

suitability

molds, yeasts

suitability

molds, yeasts

suitability

-

suitability

-

technique(s)

microbiological culture: suitable

technique(s)

microbiological culture: suitable

technique(s)

-

technique(s)

-

manufacturer/tradename

GranuCult® plus

manufacturer/tradename

NutriSelect® Basic

manufacturer/tradename

-

manufacturer/tradename

-

storage temp.

15-25°C

storage temp.

-

storage temp.

15-25°C

storage temp.

15-25°C


Still not finding the right product?

Explore all of our products under Malt extract broth


Storage Class

11 - Combustible Solids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable



Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library





Global Trade Item Number

SKUGTIN
105397050004022536064116

Questions

Reviews

No rating value

Active Filters