Select a Size
| Size/SKU | Availability | Price |
|---|---|---|
2.5 L | Please contact Customer Service for Availability | CZK 2,570.00 |
4 L | Please contact Customer Service for Availability | CZK 4,120.00 |
About This Item
CZK 2,570.00
vapor density
3.5 (vs air)
Quality Segment
vapor pressure
40 mmHg ( 20 °C), 83 mmHg ( 37.7 °C)
product line
EMPLURA®
assay
≥85% (GC)
form
liquid
autoignition temp.
433 °F
potency
>5000 mg/kg LD50, oral (Rat), >2000 mg/kg LD50, skin (Rabbit)
expl. lim.
7 %
greener alternative product characteristics
Safer Solvents and Auxiliaries
Learn more about the Principles of Green Chemistry.
sustainability
Greener Alternative Product
dilution
(low-cost alternative, for basic applications)
refractive index
n20/D 1.387 (lit.)
bp
98 °C (lit.)
mp
−91 °C (lit.)
transition temp
flash point -4 °C
solubility
0.05 g/L
density
0.684 g/mL at 25 °C (lit.)
greener alternative category
storage temp.
2-30°C
SMILES string
CCCCCCC
InChI
1S/C7H16/c1-3-5-7-6-4-2/h3-7H2,1-2H3
InChI key
IMNFDUFMRHMDMM-UHFFFAOYSA-N
General description
Legal Information
1 of 1
This Item | |||
|---|---|---|---|
| Quality Level 200 | Quality Level 200 | Quality Level 200 | Quality Level 100 |
| assay ≥85% (GC) | assay ≥99% (GC) | assay 99% | assay ≥99.3% (GC) |
| solubility 0.05 g/L | solubility 0.05 g/L | solubility water: insoluble | solubility 0.05 g/L |
| form liquid | form liquid | form - | form liquid |
| vapor pressure 40 mmHg ( 20 °C), 83 mmHg ( 37.7 °C) | vapor pressure 40 mmHg ( 20 °C), 83 mmHg ( 37.7 °C) | vapor pressure 40 mmHg ( 20 °C), 83 mmHg ( 37.7 °C) | vapor pressure 40 mmHg ( 20 °C), 83 mmHg ( 37.7 °C) |
| autoignition temp. 433 °F | autoignition temp. 433 °F | autoignition temp. 433 °F | autoignition temp. 433 °F |
Still not finding the right product?
Explore all of our products under n-Heptane
signalword
Danger
Hazard Classifications
Aquatic Acute 1 - Aquatic Chronic 1 - Asp. Tox. 1 - Flam. Liq. 2 - Skin Irrit. 2 - STOT SE 3
target_organs
Central nervous system
Storage Class
3 - Flammable liquids
wgk
WGK 2
hcodes
flash_point_f
24.8 °F - closed cup
flash_point_c
-4 °C - closed cup
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.




