Skip to Content
Merck

Skip To

1.04207

Neutral red (C.I. 50040)

Sign In to View Organizational & Contract Pricing.

About This Item

Empirical Formula (Hill Notation):
C15H17ClN4
CAS Number:
Molecular Weight:
288.78
NACRES:
NA.21
UNSPSC Code:
41116130
MDL number:


form

solid

Quality Segment

composition

Dye, ≥90% spectrophotometric assay

pH

3.1 (20 °C, 10 g/L in H2O)

solubility

50 g/L

bulk density

350‑500 kg/m3

application(s)

diagnostic assay manufacturing
hematology
histology

storage temp.

2-30°C

InChI

1S/C15H16N4.ClH/c1-9-6-13-15(8-11(9)16)18-14-7-10(19(2)3)4-5-12(14)17-13;/h4-8H,16H2,1-3H3;1H

InChI key

PGSADBUBUOPOJS-UHFFFAOYSA-N

General description

Neutral red (C.I. 50040) - for microscopy Certistain®, is a dry staining dye that is used to prepare a staining solution, that makes target structures in hematological specimen materials evaluable for diagnostic purposes. Neutral red belongs to the group of azines. Neutral red can be used for several staining such as supravital staining of leukocytes .In histology for nuclear staining, fluorescence staining, polychrome staining or for counterstaining in Gram staining. As CERTISTAIN® dye it is chemically quality-certified according to highest standards and strict specifications.

Application

for microscopy Certistain®

Analysis Note

Dye content (spectrophotometrically): ≥ 90 %
Identity (UV/VIS-Spectrum): passes test
Absorption maximum λ max (ethanol 50 %): 540 - 543 nm
Spec. Absorptivity A 1%/1cm (λmax; 0.005 g/l; ethanol 50 %): 1420 - 1610
TLC-Test: passes test
Loss on drying (110 °C): ≤ 12 %
Suitability for microscopy: passes test

Legal Information

CERTISTAIN is a registered trademark of Merck KGaA, Darmstadt, Germany

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
N70058612511.01369
form

solid

form

powder

form

powder

form

solid

application(s)

diagnostic assay manufacturing
hematology
histology

application(s)

diagnostic assay manufacturing
hematology
histology

application(s)

diagnostic assay manufacturing
hematology
histology

application(s)

-

solubility

50 g/L

solubility

water: 10 mg/mL, dark red

solubility

water: 1 mg/mL

solubility

50 g/L

composition

Dye, ≥90% spectrophotometric assay

composition

Dye content, ≥90%

composition

Dye content, 50-60%

composition

dye content, ≥75% spectrophotometric assay

storage temp.

2-30°C

storage temp.

2-8°C

storage temp.

room temp

storage temp.

2-30°C

Quality Level

200

Quality Level

200

Quality Level

100

Quality Level

200


Still not finding the right product?

Explore all of our products under Neutral red (C.I. 50040)


Storage Class

11 - Combustible Solids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable



Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

Reviews

No rating value

Active Filters