Skip to Content
Merck

Skip To

1.04170

Erythrosine B (C.I. 45430)

Sign In to View Organizational & Contract Pricing.

About This Item

Empirical Formula (Hill Notation):
C20H6I4Na2O5
CAS Number:
Molecular Weight:
879.86
UNSPSC Code:
41116130
NACRES:
NA.21
MDL number:


form

solid

Quality Segment

potency

1840 mg/kg LD50, oral (Rat)

composition

Dye, ≥80% gravimetric

pH

6-7 (20 °C, 10 g/L in H2O)

solubility

100 g/L

bulk density

470 kg/m3

application(s)

diagnostic assay manufacturing
hematology
histology

storage temp.

2-30°C

SMILES string

[Na+].[Na+].Ic1c2c(cc(c1[O-])I)C(=C4C(=C(C(=O)C(=C4)I)I)O2)c3c(cccc3)C(=O)[O-]

InChI

1S/C20H8I4O5.2Na/c21-11-5-9-13(7-3-1-2-4-8(7)20(27)28)10-6-12(22)17(26)15(24)19(10)29-18(9)14(23)16(11)25;;/h1-6,25H,(H,27,28);;/q;2*+1/p-2

InChI key

IINNWAYUJNWZRM-UHFFFAOYSA-L

General description

Erythrosin B (C.I. 45430)for microscopy Certistain®, is a dry dye for microscopy that is often used as a counterstain, vizualising proteins, connective tissues, fibers and keratin in sample material of human origin. Depending on user requirements and personal preferences, it can be used for the preparation of the commonly used aqueous or alcoholic Erythrosin B staining solutions. As the dye belongs to the portfolio of Certistain® dyes, it is chemically analyzed according to strict specifications before release and a high staining quality, with reliable staining results is guaranteed.

Application

for microscopy Certistain®

Analysis Note

Dye content (gravimetrically): ≥ 80 %
Identity
- sodium:
passes test
- UV/VIS-Spectrum: passes test
Absorption maximum λ max (water): 524 - 527 nm
Spec. Absorptivity A 1%/1cm (λmax; 0.005 g/l; water): 930 - 1170
TLC-Test: passes test
Loss on drying (110 °C): ≤ 10 %
Suitability for microscopy: passes test

Legal Information

CERTISTAIN is a registered trademark of Merck KGaA, Darmstadt, Germany


pictograms

Exclamation markEnvironment

signalword

Warning

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Chronic 2

Storage Class

11 - Combustible Solids

wgk

WGK 1

flash_point_f

359.6 °F - closed cup

flash_point_c

182 °C - closed cup



Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

Reviews

No rating value

Active Filters