Sign In to View Organizational & Contract Pricing.
Select a Size
Change View
| Size/SKU | Availability | Price |
|---|---|---|
1 g | Please contact Customer Service for Availability | CZK 14,800.00 |
About This Item
Empirical Formula (Hill Notation):
C22H20N2
CAS Number:
Molecular Weight:
312.41
UNSPSC Code:
12352116
EC Index Number:
236-069-0
NACRES:
NA.05
MDL number:
form
solid
Quality Segment
mp
228-233 °C
bulk density
380 kg/m3
storage temp.
2-30°C
SMILES string
Nc1c2c(c(cc1C)c3c4c(c(c(c3)C)N)cccc4)cccc2
InChI
1S/C22H20N2/c1-13-11-19(15-7-3-5-9-17(15)21(13)23)20-12-14(2)22(24)18-10-6-4-8-16(18)20/h3-12H,23-24H2,1-2H3
InChI key
FTEGMIZQLGXBNA-UHFFFAOYSA-N
Application
- Bacteriocin Against MRSA: This research highlights the application of ammonium sulfate in the purification of novel bacteriocins from Lactiplantibacillus plantarum for combating methicillin-resistant Staphylococcus aureus, demonstrating its critical role in biomedical research and pharmaceutical applications (Ye et al., 2024).
Analysis Note
Appearance of solution (10 g/l; ethanol): passes test
Loss on drying (110 °C): ≤ 1.0 %
Suitability as indicator (for metal titration): passes test
Suitability for bromate determination in water and drinking water for Pharo- or NOVA-spectrophotometer: passes test
Application note: Method A or Method B
Method A:For the measurement the method 195, which is pre-calibrated in the Spectroquant Pharo-and NOVA-photometers can be
used. To increase the precision of the results a self-made blank according to the application is recommended generally.
Method B:For the measurement the pre-calibrated method 195 is not applicable. Instead of that, a self-made calibration according the
application is to perform.
Loss on drying (110 °C): ≤ 1.0 %
Suitability as indicator (for metal titration): passes test
Suitability for bromate determination in water and drinking water for Pharo- or NOVA-spectrophotometer: passes test
Application note: Method A or Method B
Method A:For the measurement the method 195, which is pre-calibrated in the Spectroquant Pharo-and NOVA-photometers can be
used. To increase the precision of the results a self-made blank according to the application is recommended generally.
Method B:For the measurement the pre-calibrated method 195 is not applicable. Instead of that, a self-made calibration according the
application is to perform.
1 of 1
This Item | |||
|---|---|---|---|
| form solid | form solid | form powder | form tablet |
| storage temp. 2-30°C | storage temp. - | storage temp. - | storage temp. 2-8°C |
| Quality Level 100 | Quality Level - | Quality Level 100 | Quality Level 200 |
| mp 228-233 °C | mp 88-90 °C (lit.) | mp - | mp ≥300 °C |
| bulk density 380 kg/m3 | bulk density - | bulk density - | bulk density - |
Storage Class
11 - Combustible Solids
wgk
WGK 3
flash_point_f
Not applicable
flash_point_c
Not applicable
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.



