Skip to Content
Merck

Skip To

1.02273

Ammonium cerium(IV) sulfate dihydrate

for analysis EMSURE® ACS

Synonym(s):

Ammonium cerium(IV) sulfate dihydrate, Cerium(IV) ammonium sulfate

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice
100 g
Please contact Customer Service for Availability
CZK 3,890.00

About This Item

Linear Formula:
(NH4)4Ce(SO4)4 · 2H2O
CAS Number:
Molecular Weight:
632.55
UNSPSC Code:
12352302
EC Index Number:
231-567-4
NACRES:
NA.21
MDL number:
Assay:
≥99.0% ((NH4)4Ce(SO4)4·2H2O basis, potentiometry)
Grade:
ACS reagent
Form:
solid

CZK 3,890.00


Please contact Customer Service for Availability
Packaging: HDPE bottle

Request a Bulk Order
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


grade

ACS reagent

Quality Segment

product line

EMSURE®

assay

≥99.0% ((NH4)4Ce(SO4)4·2H2O basis, potentiometry)

form

solid

impurities

≤0.005% Substances insoluble in diluted sulfuric acid

pH

1.2 (20 °C, 100 g/L in H2O)

bulk density

800 kg/m3

anion traces

chloride (Cl-): ≤0.001%, phosphate (PO43-): ≤0.005%

cation traces

Fe: ≤0.005%, heavy metals (as Pb): ≤0.005%

storage temp.

15-25°C

SMILES string

[Ce].[S](=O)(=O)(O)O.N.O

InChI

1S/Ce.H3N.H2O4S.H2O/c;;1-5(2,3)4;/h;1H3;(H2,1,2,3,4);1H2

InChI key

AAXMWILTYFSCCH-UHFFFAOYSA-N

Analysis Note

Assay (potentiometric, (NH4)₄Ce(SO₄)₄· 2 H₂O): ≥ 99.0 %
Substances insoluble in diluted sulfuric acid: ≤ 0.005 %
Chloride (Cl): ≤ 0.001 %
Phosphate (PO₄): ≤ 0.005 %
Heavy metals (as Pb): ≤ 0.005 %
Fe (Iron): ≤ 0.005 %

Legal Information

EMSURE is a registered trademark of Merck KGaA, Darmstadt, Germany

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
1.022761.0227422269
form

solid

form

crystalline

form

solid

form

powder or crystals

assay

≥99.0% ((NH4)4Ce(SO4)4·2H2O basis, potentiometry)

assay

≥99.0% ((NH4)2[Ce(NO3)6] basis, potentiometry)

assay

≥98.0% (Ce(SO4)2.4H2O basis, potentiometry)

assay

≥98.0% (RT)

Quality Level

300

Quality Level

300

Quality Level

300

Quality Level

200

grade

ACS reagent

grade

ACS reagent

grade

-

grade

ACS reagent, puriss. p.a.

anion traces

chloride (Cl-): ≤0.001%, phosphate (PO43-): ≤0.005%

anion traces

chloride (Cl-): ≤0.001%, phosphate, silicate, arsenate (as PO43-): ≤0.005%

anion traces

chloride (Cl-): ≤0.001%, phosphate (PO43-): ≤0.01%

anion traces

phosphate (PO43-): ≤300 mg/kg

impurities

≤0.005% Substances insoluble in diluted sulfuric acid

impurities

≤0.005% Substances insoluble in diluted sulfuric acid

impurities

≤0.005% Substances insoluble in sulfuric acid

impurities

-


Still not finding the right product?


Storage Class

13 - Non Combustible Solids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable



Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

Reviews

No rating value

Active Filters