Skip to Content
Merck

Skip To

1.02005

Charcoal activated

for the determination of AOX (shaking method)

Synonym(s):

Charcoal activated

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Pack SizeSKUAvailabilityPrice
10 g
Please contact Customer Service for Availability
CZK 4,310.00

About This Item

Empirical Formula (Hill Notation):
C
CAS Number:
Molecular Weight:
12.01
MDL number:
UNSPSC Code:
11101522
EC Index Number:
231-153-3
NACRES:
SE.11

CZK 4,310.00


Please contact Customer Service for Availability

Request a Bulk OrderRequest more information
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Quality Segment

form

solid

technique(s)

photometry: suitable

matrix

Carbon base material

mp

3550 °C

bulk density

150‑440 kg/m3

storage temp.

2-30°C

SMILES string

[C]

InChI

1S/C

InChI key

OKTJSMMVPCPJKN-UHFFFAOYSA-N

Application


  • Adsorptive Removal of Dyes and Contaminants: Activated charcoal is extensively used for the adsorption and removal of dyes and other contaminants from aqueous solutions. Recent research highlights the use of activated charcoal modified with BaFe12O19 powder for the efficient removal of methyl blue dye. This magnetic adsorbent can be easily retrieved and reused, making it a sustainable option for water purification (Polley and Bera, International Journal of Environmental Analytical Chemistry, 2023).

  • Heavy Metal Detection and Removal: Activated charcoal is utilized in composite materials for detecting and removing heavy metals from water. A notable study developed a Ni-sensor based on activated charcoal plastic membranes, showcasing the material′s effectiveness in analytical applications for environmental monitoring and pollution control (Zareh et al., Journal of Analytical Chemistry, 2022).

  • Filter Composites for Industrial Wastewater Treatment: Activated charcoal combined with banana rachis cellulose nanocrystals has been used to create filter composites for industrial wastewater treatment. These composites effectively remove dyes and heavy metals, demonstrating their potential for large-scale environmental applications and enhancing water quality (Rahman et al., International Journal of Environmental Analytical Chemistry, 2022).

  • Dental Applications: In dental research, activated charcoal combined with calcium hydroxide has been evaluated for its effect on dentin microhardness. This combination shows promise in dental treatments, particularly in enhancing the mechanical properties of dentin and offering potential benefits in restorative dentistry (Abdel Hamid, Egyptian Dental Journal, 2023).

  • Environmental Adsorbents and Sensors: Activated charcoal is also used in creating advanced environmental adsorbents and sensors. For instance, a study focused on the development of magnetically retrievable BaFe12O19-activated charcoal-chitosan composite powders for the adsorption of methyl blue dye. These materials offer high efficiency, reusability, and easy recovery, making them ideal for environmental remediation applications (Polley and Bera, International Journal of Environmental Analytical Chemistry, 2022).

Analysis Note

Iodine number: ≥ 1050 mg/g
Chloride (Cl): ≤ 0.0015 %
Particle size (< 150 µm): ≥ 70
Specific surface area (BET): about 850

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
1.022161.021861.02514
form

solid

form

solid

form

solid

form

granular

matrix

Carbon base material

matrix

-

matrix

-

matrix

-

bulk density

150‑440 kg/m3

bulk density

150‑440 kg/m3

bulk density

150‑440 kg/m3

bulk density

150‑440 kg/m3

Quality Level

100

Quality Level

100

Quality Level

200

Quality Level

200

mp

3550 °C

mp

3550 °C

mp

3550 °C

mp

3550 °C

technique(s)

photometry: suitable

technique(s)

photometry: suitable

technique(s)

-

technique(s)

-


Still not finding the right product?

Explore all of our products under Charcoal activated


Storage Class

11 - Combustible Solids

wgk

nwg

flash_point_f

Not applicable

flash_point_c

Not applicable



Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Global Trade Item Number

SKUGTIN
102005001004022536027500

Questions

Reviews

No rating value

Active Filters