Skip to Content
Merck

Skip To

1.01359

Mixed indicator 4.5

pH indicator solution, acc. to Mortimer

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Pack SizeSKUAvailabilityPrice
250 mL
Please contact Customer Service for Availability
CZK 2,050.00

About This Item

UNSPSC Code:
12352401
NACRES:
NA.21

CZK 2,050.00


Please contact Customer Service for Availability
This item is subject to Excise Duty. Please check the cart for more details.

Request a Bulk Order
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Product Name

Mixed indicator 4.5, acc. to Mortimer

Quality Level

form

liquid

pH

4.8 (20 °C in H2O)

transition temp

flash point 9 °C

density

0.93 g/cm3 at 20 °C

storage temp.

2-30°C

Analysis Note

Application test: conforms
pH-value (20 °C): 4.5 - 5.0

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
1.061301.047641.09175
pH

4.8 (20 °C in H2O)

pH

4.4-5.8 (20 °C in H2O)

pH

-

pH

5.5-6.0 (20 °C in H2O)

form

liquid

form

-

form

-

form

liquid

Quality Level

200

Quality Level

200

Quality Level

200

Quality Level

200

density

0.93 g/cm3 at 20 °C

density

0.82 g/cm3 at 20 °C

density

-

density

0.89 g/cm3 at 20 °C

transition temp

flash point 9 °C

transition temp

flash point 14 °C

transition temp

-

transition temp

flash point 23 °C

storage temp.

2-30°C

storage temp.

15-25°C

storage temp.

2-30°C

storage temp.

15-25°C


pictograms

Flame

signalword

Danger

hcodes

Hazard Classifications

Flam. Liq. 2

Storage Class

3 - Flammable liquids

wgk

WGK 1

flash_point_f

48.2 °F

flash_point_c

9 °C



Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library





Global Trade Item Number

SKUGTIN
101359025004022536019741

Questions

Reviews

No rating value

Active Filters