Skip to Content
Merck

Skip To

1.01306

Naphthol green B (C.I. 10020)

pH indicator

Synonym(s):

Naphthol green B (C.I. 10020), Green acid 1

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Pack SizeSKUAvailabilityPrice
25 g
Please contact Customer Service for Availability
CZK 1,650.00

About This Item

Empirical Formula (Hill Notation):
C30H15FeN3Na3O15S3
CAS Number:
Molecular Weight:
878.46
UNSPSC Code:
12352107
EC Index Number:
243-010-2
NACRES:
NA.21
MDL number:

CZK 1,650.00


Please contact Customer Service for Availability

Request a Bulk Order
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Product Name

Naphthol green B (C.I. 10020), indicator

Quality Segment

form

solid

loss

≤10% loss on drying, 105°C

pH

9.3 (20 °C, 10 g/L in H2O)

solubility

160 g/L

bulk density

620 kg/m3

λmax

710-720 nm in water

storage temp.

2-30°C

SMILES string

[Fe](Oc5c(c6c(cc(cc6)[S](=O)(=O)[O-])cc5)N=O)(Oc3c(c4c(cc(cc4)[S](=O)(=O)[O-])cc3)N=O)Oc1c(c2c(cc(cc2)[S](=O)(=O)[O-])cc1)N=O.[Na+].[Na+].[Na+]

InChI

1S/3C10H7NO5S.Fe.3Na/c3*12-9-4-1-6-5-7(17(14,15)16)2-3-8(6)10(9)11-13;;;;/h3*1-5,12H,(H,14,15,16);;;;/q;;;+3;3*+1/p-6

InChI key

JMXROTHPANUTOJ-UHFFFAOYSA-H

Analysis Note

Identity (UV/VIS-Spectrum): passes test
Absorption maximum λmax. (water): 710 - 720 nm
Spec. Absorptivity A 1%/1cm (λmax; 0.05g/l; water; calc. on dried substance): ≥ 100
Loss on Drying (105°C): ≤ 10 %

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
705500673778364
storage temp.

2-30°C

storage temp.

room temp

storage temp.

-

storage temp.

-

bulk density

620 kg/m3

bulk density

-

bulk density

-

bulk density

-

form

solid

form

powder

form

-

form

-

pH

9.3 (20 °C, 10 g/L in H2O)

pH

-

pH

-

pH

-

Quality Level

200

Quality Level

-

Quality Level

100

Quality Level

100

solubility

160 g/L

solubility

methanol: 0.01 g/10 mL

solubility

-

solubility

-


Storage Class

11 - Combustible Solids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable



Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Global Trade Item Number

SKUGTIN
101306002504022536866741

Questions

Reviews

No rating value

Active Filters