Skip to Content
Merck

Skip To

1.00609

Nitrite Cell Test, photometric

photometric, 1.0-90.0 mg/L (NO2-N), 3.3-295.2 mg/L (NO2-), Spectroquant®

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice
25 tests
Please contact Customer Service for Availability
CZK 3,570.00

About This Item

UNSPSC Code:
41116116
NACRES:
NA.24

CZK 3,570.00


Please contact Customer Service for Availability

Request a Bulk OrderRequest more information
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Product Name

Nitrite Cell Test, photometric, 1.0-90.0 mg/L (NO2-N), 3.3-295.2 mg/L (NO2-), Spectroquant®

product line

Spectroquant®

Quality Segment

usage

sufficient for 25 tests

specific analyte(s)

nitrite

measuring range

1.0-90.0 mg/L (NO2-N), 3.3-295.2 mg/L (NO2-)

technique(s)

photometry: suitable

application(s)

boiler and cooling water
electroplating
ground and surface water
seawater
wastewater

compatibility

for use with Spectroquant® Move 100, for use with Spectroquant® Nova 60 A, for use with Spectroquant® Prove 100 (plus), for use with Spectroquant® Prove 300 (plus), for use with Spectroquant® Prove 600 (plus)

detection method

photometric (Iron sulfate)

storage temp.

15-25°C

General description

Method: photometric 1.0 - 90.0 mg/l NO₂-N 3.3 - 295.2 mg/l NO₂⁻ Spectroquant®
This Spectroquant® Nitrite Cell Test allows the accurate quantification of the nitrite content in aqueous samples.
Method applied: In acidic solution nitrite ions react with iron(II) ethylenediammonium sulfate to form a yellow to green-brown iron(II) compound that is determined photometrically.
The Spectroquant® Cell Tests come with prefilled 16 mm round cells and all the required reagents to perform the analysis according to the instruction leaflet provided.
All Spectroquant® Cell and Reagent Test Kits are equipped with the unique Live ID (2D barcode)which allows seamless method recognition and contains essential information such as lot number, expiry date, and automatic calibration updates.

Legal Information

SPECTROQUANT is a registered trademark of Merck KGaA, Darmstadt, Germany

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
1.145471.145421.14563
detection method

photometric (Iron sulfate)

detection method

photometric (Griess’ reaction)

detection method

photometric (Nitrospectral)

detection method

photometric (2,6-Dimethylphenol)

specific analyte(s)

nitrite

specific analyte(s)

nitrite

specific analyte(s)

nitrate

specific analyte(s)

nitrate

technique(s)

photometry: suitable

technique(s)

photometry: suitable

technique(s)

photometry: suitable

technique(s)

photometry: suitable

measuring range

1.0-90.0 mg/L (NO2-N), 3.3-295.2 mg/L (NO2-)

measuring range

0.010-0.700 mg/L (NO2-N), 0.03-2.30 mg/L (NO2-)

measuring range

0.5-18.0 mg/L (NO3-N), 2.2-79.7 mg/L (NO3-)

measuring range

0.5-25.0 mg/L (NO3-N), 2.2-110.7 mg/L (NO3-)

compatibility

for use with Spectroquant® Move 100, for use with Spectroquant® Prove 100 (plus), for use with Spectroquant® Prove 600 (plus), for use with Spectroquant® Nova 60 A, for use with Spectroquant® Prove 300 (plus)

compatibility

for use with Spectroquant® Move 100, for use with Spectroquant® Nova 60 A, for use with Spectroquant® Prove 100 (plus), for use with Spectroquant® Prove 300 (plus), for use with Spectroquant® Prove 600 (plus)

compatibility

for use with Spectroquant® Move 100, for use with Spectroquant® Nova 60 A, for use with Spectroquant® Prove 100 (plus), for use with Spectroquant® Prove 300 (plus), for use with Spectroquant® Prove 600 (plus)

compatibility

for use with Spectroquant® Nova 60 A, for use with Spectroquant® Prove 100 (plus), for use with Spectroquant® Prove 300 (plus), for use with Spectroquant® Prove 600 (plus)

usage

sufficient for 25 tests

usage

sufficient for 25 tests

usage

sufficient for 25 tests

usage

sufficient for 25 tests


Still not finding the right product?

Explore all of our products under Nitrite Cell Test


pictograms

CorrosionExclamation mark

signalword

Danger

hcodes

Hazard Classifications

Acute Tox. 4 Inhalation - Eye Dam. 1 - Skin Corr. 1

Storage Class

8A - Combustible corrosive hazardous materials

wgk

WGK 2



Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

Reviews

No rating value

Active Filters