Select a Size
| Size/SKU | Availability | Price |
|---|---|---|
25 pkg | Estimated to ship onMay 25, 2026fromAreál Kühne+Nagel spol. s r.o. | CZK 7,580.00 |
About This Item
Quality Segment
IVD
for in vitro diagnostic use
pH
2.5
application(s)
clinical testing
diagnostic assay manufacturing
hematology
histology
storage temp.
15-25°C
General description
The ISOSLIDE™ Alcian blue, pH 2.5 Control Slides are prepared using sections of suitable animal material, that clearly visualize acid mucosubstances. Each package of ISOSLIDE™ control slides contains 25 slides. The tissue is applied onto the slide next to the white printed labelling field. One slide is already stained with the reference method (Alcian blue solution, pH 2.5 for microscopy, Cat. no. 101647, or Alcian Blue Staining Kit, pH 2.5, Cat. no. 132657) and is used for comparison. The 24 unstained slides are stained according to the corresponding protocol, using the Alcian blue staining, e.g. the Alcian blue staining solution or with the laboratory′s own staining method. After the staining of the control slides, the staining result is to be compared with the laboratory material and the pre-stained control slide. The ISOSLIDE™ control slides are not quality-controlled for their application with molecular pathology methods and IHC.
This product is registered as IVD and CE marked. For more details, please see instructions for use (IFU). The IFU can be downloaded from this webpage.
Application
Legal Information
1 of 1
This Item | |||
|---|---|---|---|
| application(s) clinical testing | application(s) clinical testing | application(s) clinical testing | application(s) clinical testing |
| Quality Level 400 | Quality Level 400 | Quality Level 400 | Quality Level 400 |
| storage temp. 15-25°C | storage temp. 15-25°C | storage temp. 15-25°C | storage temp. 15-25°C |
| IVD for in vitro diagnostic use | IVD for in vitro diagnostic use | IVD for in vitro diagnostic use | IVD for in vitro diagnostic use |
| pH 2.5 | pH - | pH - | pH - |
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.



