Skip to Content
Merck

Skip To

1.00207

GranuCult® prime Brilliant-green Phenol-red Lactose Sucrose agar

referenced to ISO 6579, Brilliant Green Phenol Red Lactose Sucrose Agar

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Pack SizeSKUAvailabilityPrice
500 g
Please contact Customer Service for Availability
CZK 3,750.00

About This Item

NACRES:
NA.74
UNSPSC Code:
41106213

CZK 3,750.00


Please contact Customer Service for Availability

Request more information
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


agency

ISO

Quality Segment

form

powder

technique(s)

microbiological culture: suitable

application(s)

food and beverages
pharmaceutical

suitability

selective for Salmonella spp.

General description

GranuCult® prime BPLS (Brilliant Green Phenol Red Lactose Sucrose) Agar is a nutrient rich selective medium for the isolation differentiation of Salmonella spp. from food and animal feed, water and other materials.
This culture medium contains lactose, whose degradation to acid is indicated by the pH indicator phenol red, which changes its color to yellow. The indicator exhibits a pink-red color in the alkaline range.

In this highly selective culture medium, the increased growth of accompanying microorganisms is considerably prevented by raising the concentration of brilliant green.
The growth of Salmonella is improved by the richer nutrient base, but Salmonella Typhi and Shigella spp. are largely inhibited by brilliant green.

Most Salmonellae are not able to ferment either lactose or sucrose. The sucrose contained in this medium allows identification of accompanying, weakly lactose-positive or lactose-negative, but sucrose-positive microorganisms. Phenol red serves as a pH indicator and yields a yellow color as a result of in the fermentation of lactose and/or sucrose in the medium. Agar is the solidifying agent.

Application

GranuCult® prime BPLS (Brilliant Green Phenol Red Lactose Sucrose) Agar is used to isolate and differentiate Salmonella spp. from food and animal feed, water and other materials.

Other Notes

We offer two media types: the superior granulated GranuCult® and the cost-efficient powdered NutriSelect® culture media, depending on your needs.
The designations basic, plus, or prime are added to indicate the quality control level, from basic quality control to standard QC plus to prime for full regulatory compliance.

Legal Information

GRANUCULT is a registered trademark of Merck KGaA, Darmstadt, Germany
NutriSelect is a registered trademark of Merck KGaA, Darmstadt, Germany

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
1.107471.038531.03859
technique(s)

microbiological culture: suitable

technique(s)

microbiological culture: suitable

technique(s)

microbiological culture: suitable

technique(s)

microbiological culture: suitable

suitability

selective for Salmonella spp.

suitability

Salmonella spp. (spp.)

suitability

selective and differential for Salmonella Shigella

suitability

nonselective for

agency

ISO

agency

ISO 6579

agency

ISO 6579

agency

according to ISO

form

powder

form

granular

form

powder

form

granular

application(s)

food and beverages
pharmaceutical

application(s)

food and beverages
microbiology

application(s)

food and beverages
pharmaceutical

application(s)

food and beverages
pharmaceutical

Quality Level

200

Quality Level

200

Quality Level

200

Quality Level

200


Storage Class

11 - Combustible Solids

wgk

WGK 3



Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library






Global Trade Item Number

SKUGTIN
100207050004027269669720

Questions

Reviews

No rating value

Active Filters