Select a Size
| Pack Size | SKU | Availability | Price |
|---|---|---|---|
| 25 units | Please contact Customer Service for Availability | CZK 2,430.00 CZK 1,822.50 |
About This Item
CZK 1,822.50
List PriceCZK 2,430.00Save 25%product line
Spectroquant®
technique(s)
photometry: suitable
application(s)
aquaculture
boiler and cooling water
disinfection control
drinking water
ground and surface water
swimming pools
wastewater
detection method
photometric
storage temp.
15-25°C
General description
- 25 empty round cells (16 mm outer diameter) with bar code (for measuring range 0.03 - 6.00 mg/l Cl2)
- 1 white microspatula (for measuring range 0.010 - 1.000 mg/l Cl2)
- 1 AutoSelector (for measuring range 0.010 - 1.000 mg/l Cl2)
- 1 package insert It has to be purchased together with the single reagents 1.00086, 1.00087 (plus 1.00088 for total chlorine). More information can be found in the instruction leaflet.
Legal Information
1 of 1
This Item | |||
|---|---|---|---|
| product line Spectroquant® | product line - | product line - | product line - |
| detection method photometric | detection method photometric (DPD) | detection method photometric (DPD) | detection method photometric (DPD) |
| technique(s) photometry: suitable | technique(s) photometry: suitable | technique(s) photometry: suitable | technique(s) photometry: suitable |
| application(s) aquaculture | application(s) aquaculture | application(s) aquaculture | application(s) aquaculture |
| storage temp. 15-25°C | storage temp. 15-25°C | storage temp. 15-25°C | storage temp. 15-25°C |
Storage Class
10-13 - German Storage Class 10 to 13
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Global Trade Item Number
| SKU | GTIN |
|---|---|
| 1000890001 | 04027269684464 |

