Skip to Content
Merck

Skip To

0798

BCYE Agar without Cystine

plate size 90 mm, ready-to-use, sterile, pack of 20 plates, selective for bacteria: Legionella spp

Synonym(s):

Legionella Growth Medium

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Pack SizeSKUAvailabilityPrice
20 each
Check Cart for Availability
CZK 1,920.00

About This Item

UNSPSC Code:
41106217
NACRES:
NA.74
Feature:
ready-to-use
Sterility:
sterile

CZK 1,920.00


Check Cart for Availability

Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


agency

ISO 11731

sterility

sterile

form

plate (Media Filled)

feature

ready-to-use

packaging

pack of 20 plates

technique(s)

microbiological culture: suitable

plate size

90 mm

pH

6.9 ± 0.2 (25 °C)

application(s)

food and beverages
water monitoring

storage temp.

2-8°C

suitability

selective for bacteria: Legionella spp

General description

BCYE Agar without Cystine, also known as Buffered Charcoal Yeast Extract Agar, is a Ready-to-Use media designed as per ISO Standards 11731 to detect Legionella spp.

The media contains charcoal, which serves as a detoxifying agent. Yeast extract provides a rich source of vitamins, nitrogen, and carbon. ACES Buffer helps maintain an optimal pH for growth, while ferric pyrophosphate, and α-ketoglutarate stimulate the growth of Legionella species.
Legionella a gram-negative, rod-shaped bacteria, are commonly found in various freshwater biotopes, often associated with protozoans such as amoebae, where they can grow intracellularly.

Application

Buffered Charcoal Yeast Extract Agar is used to enumerate, detect and isolate Legionella spp in water samples from food and beverage industry.




Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Global Trade Item Number

SKUGTIN
79804065271834118