Skip to Content
Merck

Skip To

07-2173

Anti-Stat3 Antibody

from rabbit

Synonym(s):

Acute-phase response factor, DNA-binding protein APRF, signal transducer and activator of transcription 3, signal transducer and activator of transcription 3, (acute-phase response factor)

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice
100 μg

Estimated to ship onJune 02, 2026fromAreál Kühne+Nagel spol. s r.o.

CZK 11,000.00

About This Item

eCl@ss:
32160702
UNSPSC Code:
12352203
NACRES:
NA.41
Conjugate:
unconjugated
Clone:
polyclonal
Application:
IP, WB
Citations:
-

CZK 11,000.00


Estimated to ship onJune 02, 2026Details


Request a Bulk Order
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


biological source

rabbit

Quality Segment

conjugate

unconjugated

antibody product type

primary antibodies

clone

polyclonal

species reactivity

rat, pig, human, mouse

species reactivity (predicted by homology)

primate (based on 100% sequence homology), bovine (based on 100% sequence homology), porcine (based on 100% sequence homology)

technique(s)

immunoprecipitation (IP): suitable, western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... STAT3(6774)

General description

This protein is a member of the family of STAT proteins (Signal Transduction and Activators of Transcription). These proteins are latent cytoplasmic transcription factors with the dual function of signal transduction and activation of transcription. STATs are activated by tyrosine phosphorylation in response to different ligands, after which they translocate to the cell nucleus. The N-terminal region is highly homologous among the STAT proteins and surrounds a completely conserved arginine residue. The Stat3 protein is expressed in most cell types and is activated by multiple cytokines and growth factors including: IFN-a, IL-10, IL-6, IL-11, onM, LIF, CNTF, G-CSF, IL-12, IL-2, GH, EGF, PDGF, and CSF-1. Stat3 is not activated by IFN beta. One active Stat3 translocates to the nucleus and binds to DNA as either a homodimer or as a heterodimer with Stat1. Transcription factors associated with Stat3 include cJun and cyclic AMP responsive enhancer binding protein (CREB). Stat3 interacts with the APRE (acute phase response element) and the SIE (c-sis inducible element). These proteins are latent cytoplasmic transcription factors with the dual function of signal transduction and activation of transcription. STATs are activated by tyrosine phosphorylation in response to different ligands, after which they translocate to the cell nucleus. The N-terminal region is highly homologous among the STAT proteins and surrounds a completely conserved arginine residue.

~ 88 kDa observed.

Immunogen

Epitope: Near C-terminus
KLH-conjugated linear peptide corresponding to human STAT3 near the C-terminus.

Application

Research Category
Epigenetics & Nuclear Function
Research Sub Category
Transcription Factors
Use Anti-Stat3 Antibody (rabbit polyclonal antibody) validated in WB, IP to detect Stat3 also known as Acute-phase response factor, DNA-binding protein APRF, signal transducer & activator of transcription 3.
Western Blot (SNAP ID) Analysis: 5 µg/mL from a representative lot detected STAT3 on 10 µg of HeLa cell lysate.

Immunoprecipitation Analysis: 10 µg from a representative lot immunoprecipitated STAT3 from 500 µg of HeLa RIPA lysate.

Biochem/physiol Actions

This antibody recognizes STAT3 near the C-terminus.

Physical form

Format: Purified
Protein A
Purified rabbit polyclonal in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.

Preparation Note

Stable for 1 year at 2-8°C from date of receipt.

Analysis Note

Control
HeLa cell lysate
Evaluated by Western Blot in HeLa cell lysate.

Western Blot Analysis: 0.5 µg/mL of this antibody detected STAT3 on 10 µg of HeLa cell lysate.

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.
Replaces: AB3162

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
06-59605-107807-1347
clone

polyclonal

clone

polyclonal

clone

18F7.1, monoclonal

clone

polyclonal

biological source

rabbit

biological source

rabbit

biological source

mouse

biological source

rabbit

conjugate

unconjugated

conjugate

unconjugated

conjugate

unconjugated

conjugate

-

species reactivity

rat, pig, human, mouse

species reactivity

mouse, human, rat

species reactivity

mouse, human

species reactivity

pig, human

Gene Information

human ... STAT3(6774)

Gene Information

human ... STAT3(6774)
mouse ... Stat3(20848)

Gene Information

human ... STAT3(6774)
mouse ... Stat3(20848)

Gene Information

human ... STAT3(6774)

technique(s)

immunoprecipitation (IP): suitable, western blot: suitable

technique(s)

ChIP: suitable, electrophoretic mobility shift assay: suitable, immunocytochemistry: suitable, immunoprecipitation (IP): suitable, western blot: suitable

technique(s)

immunocytochemistry: suitable, western blot: suitable

technique(s)

western blot: suitable


Still not finding the right product?

Explore all of our products under Anti-Stat3 Antibody

or

Try our Product Selector Tool to narrow your options


Storage Class

12 - Non Combustible Liquids

wgk

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable



Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

Reviews

No rating value

Active Filters