Select a Size
| Size/SKU | Availability | Price |
|---|---|---|
100 μg | Estimated to ship onJune 05, 2026fromAreál Kühne+Nagel spol. s r.o. | CZK 14,300.00 |
About This Item
biological source
rabbit
Quality Segment
conjugate
unconjugated
antibody form
purified antibody
antibody product type
primary antibodies
clone
polyclonal
species reactivity
human, mouse
technique(s)
ELISA: suitable, immunocytochemistry: suitable, western blot: suitable
NCBI accession no.
UniProt accession no.
shipped in
wet ice
target post-translational modification
unmodified
Gene Information
human ... JMJD4(65094)
General description
JMJD4 is a transcription factor that is a putative histone demethylase. It is highly expressed in lymphoid (spleen) and hemopoietic (bone marrow) tissues
Immunogen
Application
This antibody was reported by an outside laboratory to be suitable for ELISA.
Immunocytochemistry:
HeLa and NIH/3T3 cells incubated with anti-JMJD4 (N-terminus) stained positively on nuclei.
Epigenetics & Nuclear Function
Histone Modifying Proteins
Chromatin Biology
Biochem/physiol Actions
Physical form
Preparation Note
Analysis Note
A431 lysate
1:250 dilution of this antibody detected JMJD4 on 10 µg of A431 lysate
Other Notes
Disclaimer
1 of 1
This Item | |||
|---|---|---|---|
| antibody form purified antibody | antibody form purified antibody | antibody form affinity isolated antibody | antibody form affinity isolated antibody |
| conjugate unconjugated | conjugate unconjugated | conjugate unconjugated | conjugate unconjugated |
| clone polyclonal | clone polyclonal | clone polyclonal | clone polyclonal |
| species reactivity human, mouse | species reactivity mouse, human | species reactivity human, mouse | species reactivity human |
| biological source rabbit | biological source rabbit | biological source rabbit | biological source rabbit |
| Gene Information human ... JMJD4(65094) | Gene Information human ... KDM6B(23135) | Gene Information human ... KDM3A(55818) | Gene Information human ... JMJD1C(221037) |
Still not finding the right product?
Try our Product Selector Tool to narrow your options
wgk
WGK 2
flash_point_f
Not applicable
flash_point_c
Not applicable
Storage Class
12 - Non Combustible Liquids
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.



