Sign In to View Organizational & Contract Pricing.
Select a Size
Change View
| Size/SKU | Availability | Price |
|---|---|---|
Estimated to ship onMay 25, 2026fromAreál Kühne+Nagel spol. s r.o. | CZK 29,200.00 |
About This Item
NACRES:
NA.84
UNSPSC Code:
12161703
CZK 29,200.00
Estimated to ship onMay 25, 2026Details
Quality Segment
storage temp.
-65 to -96°C
Legal Information
SMC is a registered trademark of Singulex
Storage Class
12 - Non Combustible Liquids
flash_point_f
Not applicable
flash_point_c
Not applicable
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.