Skip to Content
Merck
All Photos(1)

Key Documents

D-095

Supelco

2,5-Dimethoxy-4-methylphenethylamine hydrochloride solution

1.0 mg/mL in methanol (as free base), ampule of 1 mL, certified reference material, Cerilliant®

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C11H17NO2 · HCl
CAS Number:
Molecular Weight:
231.72
EC Number:
UNSPSC Code:
41116107

grade

certified reference material

form

liquid

feature

(Snap-N-Spike®)

packaging

ampule of 1 mL

manufacturer/tradename

Cerilliant®

drug control

Narcotic Licence Schedule E (Switzerland)

concentration

1.0 mg/mL in methanol (as free base)

technique(s)

gas chromatography (GC): suitable
liquid chromatography (LC): suitable

application(s)

pharmaceutical (small molecule)

format

single component solution

storage temp.

−20°C

InChI

1S/C11H17NO2.ClH/c1-8-6-11(14-3)9(4-5-12)7-10(8)13-2;/h6-7H,4-5,12H2,1-3H3;1H

InChI key

UIPCUUZBUOLEMM-UHFFFAOYSA-N

General description

2C-D, or 2,5-Dimethoxy-4-methylphenethylamine, is a psychedelic drug and stimulant. As a structural analog to 2C-B, 2C-C, and 2C-I, 2C-D is part of the 2C series of phenethylamines. The designer drug is sold under street names including 2C-D or 2C-M. This certified reference solution is suitable as a starting material for calibrators or controls in phenethylamine testing methods by GC/MS or LC/MS for applications in clinical toxicology, forensic analysis, or urine drug testing.

Legal Information

CERILLIANT is a registered trademark of Merck KGaA, Darmstadt, Germany
Snap-N-Spike is a registered trademark of Merck KGaA, Darmstadt, Germany

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Flam. Liq. 2 - STOT SE 1

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

49.5 °F

Flash Point(C)

9.7 °C


Regulatory Listings

Regulatory Listings are mainly provided for chemical products. Only limited information can be provided here for non-chemical products. No entry means none of the components are listed. It is the user’s obligation to ensure the safe and legal use of the product.

EU REACH Annex XVII (Restriction List)

CAS No.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service