Skip to Content
Merck
All Photos(1)

Key Documents

W346705

Sigma-Aldrich

cis-3-Octen-1-ol

≥95%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C8H16O
CAS Number:
Molecular Weight:
128.21
FEMA Number:
3467
EC Number:
Council of Europe no.:
10296
MDL number:
UNSPSC Code:
12164502
PubChem Substance ID:
Flavis number:
2.094

Pricing and availability is not currently available.

Assay

≥95%

contains

(may contain up to 0.10% alpha-tocopherol, synthetic as stabilzer)

refractive index

n20/D 1.447 (lit.)

bp

73 °C/0.5 mmHg (lit.)

density

0.849 g/mL at 25 °C (lit.)

Documentation

see Safety & Documentation for available documents

food allergen

no known allergens

SMILES string

[H]\C(CCO)=C(/[H])CCCC

Looking for similar products? Visit Product Comparison Guide

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

190.0 °F - closed cup

Flash Point(C)

87.78 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service