Skip to Content
Merck
All Photos(1)

Key Documents

A48806

Sigma-Aldrich

2-Amino-5-diethylaminopentane

97%

Synonym(s):

N1,N1-Diethyl-1,4-pentanediamine

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(C2H5)2N(CH2)3CH(NH2)CH3
CAS Number:
Molecular Weight:
158.28
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

97%

form

liquid

refractive index

n20/D 1.4429 (lit.)

density

0.817 g/mL at 25 °C (lit.)

SMILES string

CCN(CC)CCCC(C)N

InChI

1S/C9H22N2/c1-4-11(5-2)8-6-7-9(3)10/h9H,4-8,10H2,1-3H3

InChI key

CAPCBAYULRXQAN-UHFFFAOYSA-N

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Skin Corr. 1B

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 1

Flash Point(F)

154.4 °F - closed cup

Flash Point(C)

68 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Katy N Olafson et al.
Chemistry (Weinheim an der Bergstrasse, Germany), 23(55), 13638-13647 (2017-08-24)
A versatile approach to control crystallization involves the use of modifiers, which are additives that interact with crystal surfaces and alter their growth rates. Elucidating a modifier's binding specificity to anisotropic crystal surfaces is a ubiquitous challenge that is critical

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service