Skip to Content
Merck
All Photos(1)

Key Documents

862304

Sigma-Aldrich

3,4-Dehydro-L-proline

98%

Synonym(s):

(S)-3-Pyrroline-2-carboxylic acid, 3,4-Didehydro-L-proline

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C5H7NO2
CAS Number:
Molecular Weight:
113.11
Beilstein:
5376764
EC Number:
MDL number:
UNSPSC Code:
12352103
Pricing and availability is not currently available.

Assay

98%

optical activity

[α]20/D −385°, c = 2 in H2O

mp

248-250 °C (lit.)

SMILES string

OC(=O)[C@H]1NCC=C1

replaced by

Product No.
Description
Pricing

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Hyuk Sung Kwon et al.
Bioorganic & medicinal chemistry letters, 21(14), 4325-4328 (2011-06-15)
Oxygen dependent degradation of hypoxia-inducible factor (HIF)-1α is triggered with hydroxylation by proline hydroxylase domain 2 (PHD2) under normoxic conditions. Some of previously developed PHD2 inhibitors show a considerable potency against factor inhibiting HIF (FIH), the HIF asparagine hydroxylase. For
Griet Van Zeebroeck et al.
Nature chemical biology, 5(1), 45-52 (2008-12-09)
Transporter-related nutrient sensors, called transceptors, mediate nutrient activation of signaling pathways through the plasma membrane. The mechanism of action of transporting and nontransporting transceptors is unknown. We have screened 319 amino acid analogs to identify compounds that act on Gap1

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service