Skip to Content
Merck

Skip To

860239

(S)-(−)-Methioninol

98%

Synonym(s):

(S)-(−)-2-Amino-4-methylthio-1-butanol, (S)-2-Amino-4-methylmercapto-1-butanol, L-Methioninol

Sign In to View Organizational & Contract Pricing.

About This Item

Linear Formula:
CH3SCH2CH2CH(NH2)CH2OH
CAS Number:
Molecular Weight:
135.23
MDL number:
EC Number:
220-788-1
UNSPSC Code:
12352200
PubChem Substance ID:
Beilstein/REAXYS Number:
2231656


assay

98%

form

crystals

optical activity

[α]21/D −12.7°, c = 1.4 in ethanol

reaction suitability

reaction type: solution phase peptide synthesis

refractive index

n20/D 1.5216 (lit.)

application(s)

peptide synthesis

SMILES string

CSCC[C@H](N)CO

InChI

1S/C5H13NOS/c1-8-3-2-5(6)4-7/h5,7H,2-4,6H2,1H3/t5-/m0/s1

InChI key

MIQJGZAEWQQAPN-YFKPBYRVSA-N



Storage Class

11 - Combustible Solids

wgk

WGK 3

flash_point_f

235.4 °F - closed cup

flash_point_c

113 °C - closed cup

ppe

Eyeshields, Gloves, type N95 (US)



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Danielle V Miller et al.
Microbiology (Reading, England), 164(7), 969-981 (2018-06-08)
The methionine salvage pathway (MSP) is critical for regeneration of S-adenosyl-l-methionine (SAM), a widely used cofactor involved in many essential metabolic reactions. The MSP has been completely elucidated in aerobic organisms, and found to rely on molecular oxygen. Since anaerobic



Global Trade Item Number

SKUGTIN
860239-1G04061826701799

Reviews

No rating value

Active Filters