Skip to Content
Merck
All Photos(1)

Documents

682314

Sigma-Aldrich

(S)-N-Methyl-N-diphenylphosphino-1-[(R)-2-(diphenylphosphino)ferrocenyl]ethylamine

Synonym(s):

(1S)-1-(Diphenylphosphino)-2-[(1S)-1-[(diphenylphosphino)methylamino]ethyl]ferrocene, Methyl-BoPhoz

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C37H35FeNP2
CAS Number:
Molecular Weight:
611.47
MDL number:
UNSPSC Code:
12352005
PubChem Substance ID:

form

solid

mp

104.1-109.2 °C

SMILES string

[Fe].[CH]1[CH][CH][CH][CH]1.C[C@@H]([C]2[CH][CH][CH][C]2P(c3ccccc3)c4ccccc4)N(C)P(c5ccccc5)c6ccccc6

InChI

1S/C32H30NP2.C5H5.Fe/c1-26(33(2)35(29-20-11-5-12-21-29)30-22-13-6-14-23-30)31-24-15-25-32(31)34(27-16-7-3-8-17-27)28-18-9-4-10-19-28;1-2-4-5-3-1;/h3-26H,1-2H3;1-5H;/t26-;;/m1../s1

InChI key

ZIAZHUWEJYUGEO-FBHGDYMESA-N

Application

Methyl-BoPhoz can be used:
  • As an efficient ligand for asymmetric hydrogenation reactions.
  • To synthesize a single enantiomer of cyclopropylalanine via hydrogenation reaction using rhodium catalyst.
  • To prepare chiral phenylalanine derivatives from α-amido cinnamic acids (or esters) using rhodium catalyst.

Legal Information

Sold in collaboration with Johnson Matthey Catalyst for research purposes only. US6590115 and any patents arising therefrom apply.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Protecting-group-free synthesis of a dual CCK1/CCK2 receptor antagonist
Liu J, et al.
Organic & Biomolecular Chemistry, 9(8), 2654-2660 (2011)
Enantioselective Access to Chiral Drugs by using Asymmetric Hydrogenation Catalyzed by Rh (P-OP) Complexes
Etayo P, et al.
Chemistry?A European Journal , 17(50), 13978-13982 (2011)
The preparation of enantiomerically pure cyclopropylalanine.
Boaz NW, et al.
Tetrahedron Asymmetry, 14(22), 3575-3580 (2003)

Articles

P-Phos ligand family enhances catalysis, featuring atropisomeric biaryl bisphosphine with unique structural elements.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service