Sign In to View Organizational & Contract Pricing.
About This Item
Empirical Formula (Hill Notation):
C8H5N3O4
CAS Number:
Molecular Weight:
207.14
NACRES:
NA.22
PubChem Substance ID:
UNSPSC Code:
12352100
MDL number:
Assay:
96%
Form:
solid
assay
96%
form
solid
mp
345-347 °C (lit.)
functional group
nitro
SMILES string
Oc1nc2ccc(cc2nc1O)[N+]([O-])=O
InChI
1S/C8H5N3O4/c12-7-8(13)10-6-3-4(11(14)15)1-2-5(6)9-7/h1-3H,(H,9,12)(H,10,13)
InChI key
RYMLSFWVYNAKAR-UHFFFAOYSA-N
Gene Information
rat ... Gria1(50592), Grik1(29559), Grin2a(24409)
signalword
Warning
hcodes
pcodes
Hazard Classifications
Eye Irrit. 2
Storage Class
11 - Combustible Solids
wgk
WGK 3
flash_point_f
Not applicable
flash_point_c
Not applicable
ppe
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Global Trade Item Number
| SKU | GTIN |
|---|---|
| 657239-10G | 04061833432570 |
| 657239-1G | 04061826699249 |
