Skip to Content
Merck
All Photos(1)

Key Documents

545031

Sigma-Aldrich

Potassium tetracyanonickelate(II)

>99.9% trace metals basis

Synonym(s):

Dipotassium tetracyanonickelate, Potassium nickel(II) cyanide

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
K2Ni(CN)4
CAS Number:
Molecular Weight:
240.96
EC Number:
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:

Assay

>99.9% trace metals basis

form

powder

reaction suitability

reagent type: catalyst
core: nickel

SMILES string

[K+].[K+].N#C[Ni--](C#N)(C#N)C#N

InChI

1S/4CN.2K.Ni/c4*1-2;;;/q;;;;2*+1;-2

InChI key

QOOHKAMDWOKKBR-UHFFFAOYSA-N

Pictograms

Skull and crossbonesEnvironment

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 1 Dermal - Acute Tox. 2 Inhalation - Acute Tox. 2 Oral - Aquatic Acute 1 - Aquatic Chronic 1

Supplementary Hazards

Storage Class Code

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

J Silva-Avalos et al.
Applied and environmental microbiology, 56(12), 3664-3670 (1990-12-01)
Ten bacterial isolates capable of growth on tetracyanonickelate(II) [K2[Ni(CN)4]] (TCN) as the sole nitrogen source were isolated from soil, freshwater, and sewage sludge enrichments. Seven of the 10 were identified as pseudomonads, while the remaining 3 were classified as Klebsiella
S F Chow et al.
The Journal of biological chemistry, 261(16), 7264-7269 (1986-06-05)
The structure of the rhodanese-tetracyanonickelate (E X Ni(CN)2-4) complex has been characterized here in spectral and physical studies using urea as a structural perturbant. UV difference absorption, sedimentation velocity ultracentrifugation, fluorescence, and circular dichroism data show no significant conformational differences
Fehmi Bardak et al.
Spectrochimica acta. Part A, Molecular and biomolecular spectroscopy, 71(5), 1896-1900 (2008-08-23)
In this study the M(IN)(2)Ni(CN)(4) [where M: Co, Ni, and Cd, and IN: isonicotinic acid, abbreviated to M-Ni-IN] tetracyanonickelate and some metal halide complexes with the following stoichiometries: M(IN)(6)X(2) (M: Co; X: Cl and Br, and M: Ni; X: Cl
Hyouk K Kwon et al.
FEMS microbiology letters, 214(2), 211-216 (2002-09-28)
A new yeast strain capable of degrading free and metallocyanides was isolated from coke-plant wastewater. The isolated strain designated MCN2 was identified as Cryptococcus humicolus by 26S rDNA sequencing and phylogenetic analysis. During growth of the isolate with KCN as
C M Kao et al.
Letters in applied microbiology, 41(2), 216-220 (2005-07-22)
The ability of Azotobacter vinelandii, a N(2)-fixing bacterium, to biodegrade tetracyanonickelate (TCN) was evaluated. The amounts of TCN were measured spectrophotometrically. Ammonia was determined colorimetrically by the indophenol method. The produced methane from TCN conversion by A. vinelandii was detected

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service