Skip to Content
Merck

Skip To

RPMI 8866 Cell Line human

NOTE: Both the cell line and DNA from the cell line may be available for this product. Please choose -1VL or VIAL for cells, or -DNA-5UG for DNA, 95041316, human blood, Lymphoblastoid

Synonym(s):

R8866 Cells, RPMI8866 Cells, Roswell Park Memorial Institute 8866 Cells

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice

About This Item

UNSPSC Code:
41106514
Biological source:
human blood
Relevant disease(s):
cancer
Growth mode:
Suspension
Karyotype:
Not specified
Morphology:
Lymphoblastoid
Manufacturer/tradename:
ECACC
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


biological source

human blood

manufacturer/tradename

ECACC

packaging

tube of 5 μg 95041316-DNA-5UG, pkg of vial of cells 95041316-1VL

growth mode

Suspension

karyotype

Not specified

morphology

Lymphoblastoid

products

Not specified

receptors

Immunoglobulin E (IgE) binding factor, Calcitonin

technique(s)

cell culture | mammalian: suitable

relevant disease(s)

cancer

shipped in

dry ice

storage temp.

−196°C

Application

Receptor studies, binding assays, hormone studies.

Biochem/physiol Actions

Human B lymphocyte
RPMI 8866 has been established from the peripheral blood of a 51-year-old American woman with chronic myelogenous leukaemia in May 1966. The B lymphoid cell line expresses a relatively high amount of calcitonin receptors, membrane bound Ig and soluble IgE-binding factor (Fc epsilon RII/CD23).
STR-PCR Data: Amelogenin: X
CSF1PO: 11
D13S317: 10,14
D16S539: 11
D5S818: 12
D7S820: 11,12
THO1: 9.3
TPOX: 8,11
vWA: 14,16

Preparation Note

RPMI 1640 + 2mM Glutamine + 10% Foetal Bovine Serum (FBS).
To keep the cells in exponential growth maintain cultures between 3-9x100,000 cells/ml; 5% CO2; 37°C. If starting from a frozen ampoule, add thawed cells to a conical based centrifuge tube e.g. 15ml tube. Slowly add 4 ml of culture medium to the tube. Take a sample of the cell suspension, e.g. 200ul, to count cells. Centrifuge the cell suspension at low speed i.e. 100 - 150 x g for a maximum of 5 minutes. Remove medium and re-suspend the cell pellet at a density of 3 - 5 x 100,000 cells/ml in fresh medium containing 10% serum. Incubate flask at 37 C; 5 - 7% CO2. Check daily. Keep flask in a vertical position until the cells reach the exponential phase of growth, this can take up to 7 days.

Other Notes

Additional freight & handling charges may be applicable for Asia-Pacific shipments. Please check with your local Customer Service representative for more information.
This cell line is part of the European Collection of Authenticated Cell Cultures (ECACC), an international repository managed by the United Kingdom Health Security Agency (UKHSA). No licensing agreement is required when either this cell line or the DNA extracted from it are used for internal research purposes only. Any other use of these products is prohibited without the express written permission of UKHSA. Inquiries regarding authorized use of this cell line or its genomic DNA may be directed to culturecollections@ukhsa.gov.uk.

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
CB_87012702CB_91062702CB_91051511
vibrant-m

RPMI 8226

description

NOTE: Both the cell line and DNA from the cell line may be available for this product. Please choose -1VL or VIAL for cells, or -DNA-5UG for DNA, 95041316, human blood, Lymphoblastoid

description

NOTE: Both the cell line and DNA from the cell line may be available for this product. Please choose -1VL or VIAL for cells, or -DNA-5UG for DNA, 87012702, human blood, Not specified

description

NOTE: Both the cell line and DNA from the cell line may be available for this product. Please choose -1VL or VIAL for cells, or -DNA-5UG for DNA. 91062702, mouse blood, Macrophage

description

NOTE: Both the cell line and DNA from the cell line may be available for this product. Please choose -1VL or VIAL for cells, or -DNA-5UG for DNA, 91051511, mouse blood, Not specified

growth mode

Suspension

growth mode

Suspension

growth mode

Semi-adherent

growth mode

Semi-adherent

karyotype

Not specified

karyotype

Not specified

karyotype

Not specified

karyotype

Not specified

shipped in

dry ice

shipped in

dry ice

shipped in

dry ice

shipped in

dry ice

packaging

tube of 5 μg 95041316-DNA-5UG, pkg of vial of cells 95041316-1VL

packaging

pkg of vial of cells 87012702-1VL, tube of 5 μg 87012702-DNA-5UG

packaging

tube of 5 μg 91062702-DNA-5UG, vial of  cells 91062702-1VL

packaging

pkg of vial of cells 91051511-1VL, tube of 5 μg 91051511-DNA-5UG

biological source

human blood

biological source

human blood

biological source

mouse blood

biological source

mouse blood

storage temp.

−196°C

storage temp.

−196°C

storage temp.

−196°C

storage temp.

−196°C




Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

1–8 of 8 Questions  
  1. Could you provide information on product 95041316-1VL, including the doubling time and data sheet details?

    1 answer
    1. The RPMI 8866 cell line datasheet is accessible through the cell line catalogue entry on the website. Our ECACC cell culturists have recommended that this cell line might require up to 7 days to reach the exponential or log phase of the cell cycle. Once it reaches this phase, the doubling time of the cell line is expected to be around 18-24 hours, provided the cells are seeded at the optimal subculture seeding density of between 2-3 x 10^4 cells/ml.

      Helpful?

  2. How many Cell Line cells are in the vial?

    1 answer
    1. Each vial of cells contains 2-3 x 106 cells in 1 ml of freezing media. This is in a 1.8 mL ampule.  For exceptionally large cells, counts may decrease. Suspension cells, generally smaller cells, may contain as many as 4-5 x 106 cells/vial to assure optimal viability upon thaw.

      Helpful?

  3. What passage are my Cell Line cells?

    1 answer
    1. If the passage of the cells is known, it is listed on the product page at the HPA website (www.hpacultures.org.uk).

      Helpful?

  4. How do I handle Cell Line frozen cells upon arrival?

    1 answer
    1. Upon receipt, frozen ampules should be transferred directly to vapor phase liquid nitrogen without delay (-135°C) or liquid phase liquid nitrogen if vapor phase is not available. DO NOT use a -80°C freezer as an alternative; this will result in loss of viability.

      Helpful?

  5. Are my Cell Line cells mycoplasma tested?

    1 answer
    1. ECACC routinely tests all manufactured cell banks for mycoplasma.

      Helpful?

  6. Why is Vapor Phase Liquid Nitrogen preferred for storage of Cell Line cells?

    1 answer
    1. If ampules are immersed into liquid phase of liquid nitrogen, it increases the risk of the liquid seeping into the vial. This could lead to problems of cross-contamination and increased risk of the ampule exploding when thawed.

      Helpful?

  7. Are the Cell Line cells tested for viral pathogens?

    1 answer
    1. HPA Cultures does not perform any viral testing on the cell lines.

      Helpful?

  8. What medium should I use when I thaw the Cell Line cells?

    1 answer
    1. The medium for each cell line is listed on the product page. It is specific to each cell line. If not visible on the Sigma-Aldrich website product page, the HPA website also contains the same information (www.hpacultures.org.uk). The product number for the cell line is the same on either site.

      Helpful?

Reviews

No rating value

Active Filters