Skip to Content
Merck
All Photos(1)

Documents

424846

Sigma-Aldrich

Benzyl (S)-(−)-lactate

technical grade, 90%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3CH(OH)CO2CH2C6H5
CAS Number:
Molecular Weight:
180.20
MDL number:
UNSPSC Code:
12352108
PubChem Substance ID:

grade

technical grade

Assay

90%

form

liquid

optical activity

[α]20/D −19°, c = 1 in acetone

refractive index

n20/D 1.511 (lit.)

bp

105-109 °C/0.1 mmHg (lit.)

density

1.12 g/mL at 25 °C (lit.)

SMILES string

C[C@H](O)C(=O)OCc1ccccc1

InChI

1S/C10H12O3/c1-8(11)10(12)13-7-9-5-3-2-4-6-9/h2-6,8,11H,7H2,1H3/t8-/m0/s1

InChI key

ZYTLPUIDJRKAAM-QMMMGPOBSA-N

General description

Benzyl (S)-(-)-lactate may be used in the preparation of (R)-(+)-methyl 7-(1-(benzyloxy)-1-oxopropan-2-yloxy)-2-oxo-2H-chromene-3-carboxylate.

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Synthesis and evaluation of two coumarin-type derivatization reagents for fluorescence detection of chiral amines and chiral carboxylic acids.
Mertens MD and Gutschow M.
Chirality, 25(12), 957-964 (2013)
Bunta Watanabe et al.
PloS one, 10(8), e0136242-e0136242 (2015-08-22)
CCG-1423 suppresses several pathological processes including cancer cell migration, tissue fibrosis, and the development of atherosclerotic lesions. These suppressions are caused by inhibition of myocardin-related transcription factor A (MRTF-A), which is a critical factor for epithelial-mesenchymal transition (EMT). CCG-1423 can

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service