358754
1,3,4,6,7,8-Hexahydro-2H-pyrimido[1,2-a]pyrimidine, polymer-bound
200-400 mesh, extent of labeling: 2.6 mmol/g loading, 2 % cross-linked with divinylbenzene
Synonym(s):
1,3,4,6,7,8-Hexahydro-2H-pyrimido[1,2-a]pyrimidine bound to polystyrene, 1,5,7-Triazabicyclo[4.4.0]dec-5-ene–polystyrene crosslinked with 2% DVB, ‘7-Methyl-1,5,7-triazabicyclo[4.4.0]dec-5-ene’ on polystyrene
About This Item
Recommended Products
crosslinking
2 % cross-linked with divinylbenzene
reaction suitability
reaction type: solution phase peptide synthesis
reactivity: proton reactive
extent of labeling
2.6 mmol/g loading
particle size
200-400 mesh
Looking for similar products? Visit Product Comparison Guide
Related Categories
Features and Benefits
Signal Word
Danger
Hazard Statements
Precautionary Statements
Hazard Classifications
Skin Corr. 1B
Storage Class Code
8A - Combustible corrosive hazardous materials
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Personal Protective Equipment
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service