Skip to Content
Merck
All Photos(1)

Documents

408530

Sigma-Aldrich

Z-L-Isoleucine

98%

Synonym(s):

N-Carbobenzyloxy-L-isoleucine, Z-Ile-OH

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3CH2CH(CH3)CH(NHCOOCH2C6H5)COOH
CAS Number:
Molecular Weight:
265.30
Beilstein:
4189486
EC Number:
MDL number:
UNSPSC Code:
12352200
eCl@ss:
32160406
PubChem Substance ID:
NACRES:
NA.22

Assay

98%

form

solid

optical activity

[α]20/D +6.0°, c = 6 in ethanol

reaction suitability

reaction type: solution phase peptide synthesis

mp

52-54 °C (lit.)

application(s)

peptide synthesis

SMILES string

CCC(C)[C@H](NC(=O)OCc1ccccc1)C(O)=O

InChI

1S/C14H19NO4/c1-3-10(2)12(13(16)17)15-14(18)19-9-11-7-5-4-6-8-11/h4-8,10,12H,3,9H2,1-2H3,(H,15,18)(H,16,17)/t10-,12-/m0/s1

InChI key

JSHXJPFZKBRLFU-JQWIXIFHSA-N

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Griet Van Zeebroeck et al.
Nature chemical biology, 5(1), 45-52 (2008-12-09)
Transporter-related nutrient sensors, called transceptors, mediate nutrient activation of signaling pathways through the plasma membrane. The mechanism of action of transporting and nontransporting transceptors is unknown. We have screened 319 amino acid analogs to identify compounds that act on Gap1

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service