308218
2,6-Diisopropylphenyl isocyanate
technical grade, 90%
About This Item
Recommended Products
grade
technical grade
Assay
90%
refractive index
n20/D 1.519 (lit.)
bp
117 °C/10 mmHg (lit.)
density
0.951 g/mL at 25 °C (lit.)
SMILES string
CC(C)c1cccc(C(C)C)c1N=C=O
InChI
1S/C13H17NO/c1-9(2)11-6-5-7-12(10(3)4)13(11)14-8-15/h5-7,9-10H,1-4H3
InChI key
FEUFNKALUGDEMQ-UHFFFAOYSA-N
Gene Information
human ... GABRA1(2554)
Looking for similar products? Visit Product Comparison Guide
Application
Signal Word
Danger
Hazard Statements
Precautionary Statements
Hazard Classifications
Acute Tox. 1 Inhalation - Acute Tox. 4 Oral - Eye Dam. 1 - Resp. Sens. 1 - Skin Corr. 1B - STOT SE 3
Target Organs
Respiratory system
Storage Class Code
6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials
WGK
WGK 2
Flash Point(F)
224.6 °F - closed cup
Flash Point(C)
107 °C - closed cup
Personal Protective Equipment
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service