Skip to Content
Merck
All Photos(2)

Documents

277924

Sigma-Aldrich

4,4′-Methylenebis(2,6-di-tert-butylphenol)

98%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH2[C6H2[C(CH3)3]2OH]2
CAS Number:
Molecular Weight:
424.66
Beilstein:
1916919
EC Number:
MDL number:
UNSPSC Code:
12162002
PubChem Substance ID:
NACRES:
NA.23

Assay

98%

form

solid

bp

289 °C/40 mmHg (lit.)

mp

155-159 °C (lit.)

SMILES string

CC(C)(C)c1cc(Cc2cc(c(O)c(c2)C(C)(C)C)C(C)(C)C)cc(c1O)C(C)(C)C

InChI

1S/C29H44O2/c1-26(2,3)20-14-18(15-21(24(20)30)27(4,5)6)13-19-16-22(28(7,8)9)25(31)23(17-19)29(10,11)12/h14-17,30-31H,13H2,1-12H3

InChI key

MDWVSAYEQPLWMX-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Antioxidant.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

O Takahashi et al.
Toxicology letters, 8(1-2), 77-86 (1981-04-01)
Hepatic lipids were studied in Sprague-Dawley male rats given, 2,2'-methylenebis(4-ethyl-6-tert-butylphenol), 2,2'-methylenebis(4-methyl-6-tert-butylphenol), 4,4'-butylidenebis(3-methyl-6-tert-butylphenol) or 4,4'-methylenebis(2,6-di-tert-butylphenol) at a level of 1.135 mmol% for 1 week. 4,4'-Butylidenebis(3-methyl-6-tert-butylphenol) induced fatty liver and increased triglyceride, diglyceride and cholesteryl-ester concentrations, 7.1-, 5.8- and 6.1-fold, respectively. Plasma
Yanfeng Liu et al.
Toxicology in vitro : an international journal published in association with BIBRA, 52, 321-331 (2018-07-22)
The purpose of present study was to investigate the applicability of reconstructed human epidermis model to identify skin corrosive UN GHS Categories 1A, 1B/1C and non-corrosive chemicals in China. By using a commercialized reconstructed human epidermis model, China EpiSkin™ which

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service