![4,4′-Methylenebis(2,6-di-tert-butylphenol) for synthesis](/deepweb/assets/sigmaaldrich/product/images/195/862/0a03226b-fce8-4bce-a34a-ff2438dd8ddd/640/0a03226b-fce8-4bce-a34a-ff2438dd8ddd.jpg)
4,4′-Methylenebis(2,6-di-tert-butylphenol)
![4,4′-Methylenebis(2,6-di-tert-butylphenol) for synthesis](/deepweb/assets/sigmaaldrich/product/images/195/862/0a03226b-fce8-4bce-a34a-ff2438dd8ddd/640/0a03226b-fce8-4bce-a34a-ff2438dd8ddd.jpg)
Synonym(s):
4,4′-Methylenebis(2,6-di-tert-butylphenol)
Empirical Formula (Hill Notation):
C29H44O2
CAS Number:
Molecular Weight:
424.66
Products
Brand
Formula Weight
Physical Form
Quality Segment
Showing 1-1 of 1
Compare
Product Number
Description
Pricing