Skip to Content
Merck
All Photos(1)

Key Documents

208809

Sigma-Aldrich

1-Bromo-2,2-dimethoxypropane

97%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3C(OCH3)2CH2Br
CAS Number:
Molecular Weight:
183.04
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

97%

form

liquid

impurities

1-2% 3-bromo-2-methoxy-1-propene

refractive index

n20/D 1.447 (lit.)

bp

83-87 °C/80 mmHg (lit.)

density

1.355 g/mL at 25 °C (lit.)

SMILES string

COC(C)(CBr)OC

InChI

1S/C5H11BrO2/c1-5(4-6,7-2)8-3/h4H2,1-3H3

InChI key

SGTITUFGCGGICE-UHFFFAOYSA-N

Application

1-Bromo-2,2-dimethoxypropane was used in the synthesis of:
  • coronene[1]
  • dicarbonyl(η5-cyclopentadienyl)(η′-2-methoxyallyl)iron[2]

Pictograms

FlameExclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Flam. Liq. 3 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

105.8 °F - closed cup

Flash Point(C)

41 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Gram scale synthesis of Benzo [ghi] perylene and coronene.
Van Dijk JTM, et al.
The Journal of Organic Chemistry, 61(3), 1136-1139 (1996)
Synthesis of Dicarbonyl (η5-Cyclopentadienyl)(η′-2-Methoxyallyl) iron.
Abram TS and Baker R.
Synth. React. Inorg. Met.-Org. Chem. , 9(5), 471-477 (1979)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service