Skip to Content
Merck
All Photos(2)

Key Documents

150371

Sigma-Aldrich

2-Acetyl-1-tetralone

98%

Sign Into View Organizational & Contract Pricing

Select a Size

5 G
CZK 1,393.00
25 G
CZK 8,750.00

CZK 1,393.00

List PriceCZK 1,990.00Save 30%

Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
5 G
CZK 1,393.00
25 G
CZK 8,750.00

About This Item

Empirical Formula (Hill Notation):
C12H12O2
CAS Number:
Molecular Weight:
188.22
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

CZK 1,393.00

List PriceCZK 1,990.00Save 30%

Please contact Customer Service for Availability

Request a Bulk Order

Assay

98%

form

solid

bp

135-142 °C/1 mmHg (lit.)

mp

55-57 °C (lit.)

functional group

ketone

SMILES string

CC(=O)C1CCc2ccccc2C1=O

InChI

1S/C12H12O2/c1-8(13)10-7-6-9-4-2-3-5-11(9)12(10)14/h2-5,10H,6-7H2,1H3

InChI key

BSELJBOMYPMBNL-UHFFFAOYSA-N

Application

2-Acetyl-1-tetralone was used in the preparation of dihydronaphtho-pyrrole[1].

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

221.0 °F - closed cup

Flash Point(C)

105.00 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Porphyrins with exocyclic rings. Part 5. Synthesis of a naphtho [1, 2-b] porphyrin.
Lash TD and Denny CP.
Tetrahedron, 51(1), 59-66 (1995)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service