Skip to Content
Merck
All Photos(1)

Documents

299553

Sigma-Aldrich

2-Acetyl-5-methylfuran

98%

Synonym(s):

NSC 80404

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C7H8O2
CAS Number:
Molecular Weight:
124.14
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

vapor density

>1 (vs air)

Assay

98%

form

liquid

refractive index

n20/D 1.512 (lit.)

bp

100-101 °C/25 mmHg (lit.)

density

1.066 g/mL at 25 °C (lit.)

SMILES string

CC(=O)c1ccc(C)o1

InChI

1S/C7H8O2/c1-5-3-4-7(9-5)6(2)8/h3-4H,1-2H3

InChI key

KEFJLCGVTHRGAH-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

s-cis-trans isomerism of 2-acetyl-5-methylfuran was investigated by IR and NMR spectroscopy.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

176.0 °F - closed cup

Flash Point(C)

80 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Roberto Rittner et al.
Spectrochimica acta. Part A, Molecular and biomolecular spectroscopy, 103, 84-89 (2012-12-25)
The s-cis-trans isomerism of two furan derivatives [2-acetyl- (AF) and 2-acetyl-5-methylfuran, (AMF)] was analyzed, using data from the deconvolution of their carbonyl absorption band in two solvents (CH(2)Cl(2) and CH(3)CN). These infrared data showed that the O,O-trans conformers predominate in
P S Sujatha
Food and chemical toxicology : an international journal published for the British Industrial Biological Research Association, 46(1), 286-292 (2007-10-02)
Furfuryl alcohol (FA) and 2-furyl methyl ketone (2FMK) are two dietary furans with wide industrial applications and also found in a variety of food items. In a mouse test system, the mutagenicity of these two compounds after five days of
J Jodynis-Liebert
Xenobiotica; the fate of foreign compounds in biological systems, 18(8), 887-892 (1988-08-01)
1. Continuous extraction, column chromatography and t.l.c. were employed to isolate a minor metabolite of 5-methyl-2-furaldehyde from rat urine. 2. The metabolite was identified by mass spectrometry and independent synthesis as 5-methyl-2-furylmethylketone. 3. A method for quantitative determination of the

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service