Skip to Content
Merck
All Photos(1)

Key Documents

10969

Sigma-Aldrich

Butyl decanoate

≥98.0%

Synonym(s):

Butyl caprate, Capric acid butyl ester

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3(CH2)8COO(CH2)3CH3
CAS Number:
Molecular Weight:
228.37
Beilstein:
1769651
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

≥98.0%

refractive index

n20/D 1.430

density

0.86 g/mL at 20 °C (lit.)

SMILES string

CCCCCCCCCC(=O)OCCCC

InChI

1S/C14H28O2/c1-3-5-7-8-9-10-11-12-14(15)16-13-6-4-2/h3-13H2,1-2H3

InChI key

ZRNCNTSXSYXHOW-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Butyl decanoate is a colorless liquid. The IR spectra of butyl decanoate was measured in various solvents[1].

Application

Butyl decanoate poses no health hazard and is biodegradable. Hence it is used to substitute hazardous compounds used in tissue processing and dewaxing, along with other compounds without impairing the morphology, staining characteristics, or diagnostic value of the tissue sections[2]. It was used as an internal standard in the calculation of acid concentrations in the whole egg samples[3].

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Quantitative Determination of Lactic and Succinic Acids in Frozen Whole Eggs by Gas?Liquid Chromatography.
Journal of Food Science, 34(1), 37-42 (1969)
H Lyon et al.
Histochemistry and cell biology, 103(4), 263-269 (1995-04-01)
The aim of this study was to substitute hazardous compounds, used in tissue processing and dewaxing, with compounds having lowest possible toxicity and inflammability without impairing the morphology, staining characteristics, or diagnostic value of the tissue sections. All aromatic compounds
Equilibrium constant for the lipase-catalyzed synthesis of fatty acid butyl ester in various organic solvents.
Kobayashi T, et al.
Journal of Molecular Catalysis. B, Enzymatic, 24, 61-66 (2003)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service