Přejít k obsahu
Merck
  • Differential expression of the protein kinase A subunits in normal adrenal glands and adrenocortical adenomas.
Differential expression of the protein kinase A subunits in normal adrenal glands and adrenocortical adenomas.
Scientific reports (2017-03-03)
Isabel Weigand, Cristina L Ronchi, Marthe Rizk-Rabin, Guido Di Dalmazi, Vanessa Wild, Kerstin Bathon, Beatrice Rubin, Davide Calebiro, Felix Beuschlein, Jérôme Bertherat, Martin Fassnacht, Silviu Sbiera
ANOTACE
Somatic mutations in protein kinase A catalytic α subunit (PRKACA) were found to be causative for 30-40% of cortisol-producing adenomas (CPA) of the adrenal gland, rendering PKA signalling constitutively active. In its resting state, PKA is a stable and inactive heterotetramer, consisting of two catalytic and two regulatory subunits with the latter inhibiting PKA activity. The human genome encodes three different PKA catalytic subunits and four different regulatory subunits that are preferentially expressed in different organs. In normal adrenal glands all regulatory subunits are expressed, while CPA exhibit reduced protein levels of the regulatory subunit IIβ. In this study, we linked for the first time the loss of RIIβ protein levels to the PRKACA mutation status and found the down-regulation of RIIβ to arise post-transcriptionally. We further found the PKA subunit expression pattern of different tumours is also present in the zones of the normal adrenal cortex and demonstrate that the different PKA subunits have a differential expression pattern in each zone of the normal adrenal gland, indicating potential specific roles of these subunits in the regulation of different hormones secretion.
MATERIÁLY
Číslo produktu
Značka
Popis produktu
Sigma-Aldrich
Anti-PRKAR2B antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution