Přejít k obsahu
Merck
  • Enhanced aerobic exercise performance in women by a combination of three mineral Chelates plus two conditionally essential nutrients.
Enhanced aerobic exercise performance in women by a combination of three mineral Chelates plus two conditionally essential nutrients.
Journal of the International Society of Sports Nutrition (2017-11-22)
Robert A DiSilvestro, Staci Hart, Trisha Marshall, Elizabeth Joseph, Alyssa Reau, Carmen B Swain, Jason Diehl
ANOTACE
Certain essential and conditionally essential nutrients (CENs) perform functions involved in aerobic exercise performance. However, increased intake of such nutrient combinations has not actually been shown to improve such performance. For 1 mo, aerobically fit, young adult women took either a combination of 3 mineral glycinate complexes (daily dose: 36 mg iron, 15 mg zinc, and 2 mg copper) + 2 CENs (daily dose: 2 g carnitine and 400 mg phosphatidylserine), or the same combination with generic mineral complexes, or placebo ( In Trial 1, the mineral glycinates + CENs decreased 3 mile run time (25.6 ± 2.4 vs 26.5 ± 2.3 min, In summary, a combination of certain mineral complexes plus two CENs improved aerobic exercise performance in fit young adult women.
MATERIÁLY
Číslo produktu
Značka
Popis produktu
Sigma-Aldrich
Copper(II) D-gluconate, 98%