Přejít k obsahu
Merck
  • Study of the simulated sunlight photolysis mechanism of ketoprofen: the role of superoxide anion radicals, transformation byproducts, and ecotoxicity assessment.
Study of the simulated sunlight photolysis mechanism of ketoprofen: the role of superoxide anion radicals, transformation byproducts, and ecotoxicity assessment.
Environmental science. Processes & impacts (2017-07-27)
Yingfei Wang, Wen Deng, Fengliang Wang, Yuehan Su, Yiping Feng, Ping Chen, Jingshuai Ma, Haiying Su, Kun Yao, Yang Liu, Wenying Lv, Guoguang Liu
ANOTACE
The aim of this study was to investigate the photolysis mechanism of ketoprofen (KET) under simulated sunlight. The results demonstrated that the photolysis of KET aligned well with pseudo first-order kinetics. Radical scavenging experiments and dissolved oxygen experiments revealed that the superoxide anion radical (O
MATERIÁLY
Číslo produktu
Značka
Popis produktu
Sigma-Aldrich
Sodium azide, ≥99%, ultra dry