Přejít k obsahu
Merck
  • Flexible transparent displays based on core/shell upconversion nanophosphor-incorporated polymer waveguides.
Flexible transparent displays based on core/shell upconversion nanophosphor-incorporated polymer waveguides.
Scientific reports (2017-04-04)
Bong Je Park, A-Ra Hong, Suntak Park, Ki-Uk Kyung, Kwangyeol Lee, Ho Seong Jang
ANOTACE
Core/shell (C/S)-structured upconversion nanophosphor (UCNP)-incorporated polymer waveguide-based flexible transparent displays are demonstrated. Bright green- and blue-emitting Li(Gd,Y)F
MATERIÁLY
Číslo produktu
Značka
Popis produktu
Sigma-Aldrich
Tetra(ethylene glycol) diacrylate, technical grade, contains 100-150 ppm HQ as inhibitor, 150-200 ppm MEHQ as inhibitor