Přejít k obsahu
Merck
Epithelial expression and function of trypsin-3 in irritable bowel syndrome.
Gut (2017-01-18)
Claire Rolland-Fourcade, Alexandre Denadai-Souza, Carla Cirillo, Cintya Lopez, Josue Obed Jaramillo, Cleo Desormeaux, Nicolas Cenac, Jean-Paul Motta, Muriel Larauche, Yvette Taché, Pieter Vanden Berghe, Michel Neunlist, Emmanuel Coron, Sylvain Kirzin, Guillaume Portier, Delphine Bonnet, Laurent Alric, Stephen Vanner, Celine Deraison, Nathalie Vergnolle
ANOTACE
Proteases are key mediators of pain and altered enteric neuronal signalling, although the types and sources of these important intestinal mediators are unknown. We hypothesised that intestinal epithelium is a major source of trypsin-like activity in patients with IBS and this activity signals to primary afferent and enteric nerves and induces visceral hypersensitivity. Trypsin-like activity was determined in tissues from patients with IBS and in supernatants of Caco-2 cells stimulated or not. These supernatants were also applied to cultures of primary afferents. mRNA isoforms of trypsin ( We showed that stimulated intestinal epithelial cells released trypsin-like activity specifically from the basolateral side. This activity was able to activate sensory neurons. In colons of patients with IBS, increased trypsin-like activity was associated with the epithelium. We identified that trypsin-3 was the only form of trypsin upregulated in stimulated intestinal epithelial cells and in tissues from patients with IBS. Trypsin-3 was able to signal to human submucosal enteric neurons and mouse sensory neurons, and to induce visceral hypersensitivity in vivo, all by a protease-activated receptor-2-dependent mechanism. In IBS, the intestinal epithelium produces and releases the active protease trypsin-3, which is able to signal to enteric neurons and to induce visceral hypersensitivity.
MATERIÁLY
Číslo produktu
Značka
Popis produktu
Sigma-Aldrich
Nafamostat mesylate, ≥98% (HPLC)