Přejít k obsahu
Merck
  • Synthesis, radiosynthesis and in vitro evaluation of 18F-Bodipy-C16/triglyceride as a dual modal imaging agent for brown adipose tissue.
Synthesis, radiosynthesis and in vitro evaluation of 18F-Bodipy-C16/triglyceride as a dual modal imaging agent for brown adipose tissue.
PloS one (2017-08-18)
Andreas Paulus, Marco Maenen, Natascha Drude, Emmani B M Nascimento, Wouter D van Marken Lichtenbelt, Felix M Mottaghy, Matthias Bauwens
ANOTACE
Brown adipose tissue research is in the focus in the field of endocrinology. We designed a dual-modal fluorescent/PET fatty acid based tracer on commercially available Bodipy-C16, which can be synthesized to its corresponding triglyceride and which combines the benefits of fluorescent and PET imaging. Bodipy-C16 was coupled to 1,3-diolein resulting in Bodipy-triglyceride. Bodipy-C16 and Bodipy-triglyceride compounds were radiolabeled with 18F using an 18F/19F exchange reaction to yield a dual-modal imaging molecule. Uptake of radiolabeled and non-labeled Bodipy-C16 and Bodipy-triglyceride was analyzed by fluorescence imaging and radioactive uptake in cultured adipocytes derived from human brown adipose tissue and white adipose tissue. Bodipy-C16 and Bodipy-triglyceride were successfully radiolabeled and Bodipy-C16 showed high shelf life and blood plasma stability (99% from 0-4 h). The uptake of Bodipy-C16 increased over time in cultured adipocytes, which was further enhanced after beta-adrenergic stimulation with norepinephrine. The uptake of Bodipy-C16 was inhibited by oleic acid and CD36 inhibitor sulfosuccinimidyl-oleate. The poor solubility of Bodipy-triglyceride did not allow stability or in vitro experiments. The new developed dual modal fatty acid based tracers Bodipy-C16 and Bodipy-triglyceride showed promising results to stimulate further in vivo evaluation and will help to understand brown adipose tissues role in whole body energy expenditure.
MATERIÁLY
Číslo produktu
Značka
Popis produktu
Sigma-Aldrich
1,3-Diolein, ≥99% (GC)