Přejít k obsahu
Merck
  • Bioanalytical Method for the Determination of Hydroxyproline in Mouse Kidney by High-Performance Liquid Chromatography with Tandem Mass Spectrometric Detection.
Bioanalytical Method for the Determination of Hydroxyproline in Mouse Kidney by High-Performance Liquid Chromatography with Tandem Mass Spectrometric Detection.
Analytical sciences : the international journal of the Japan Society for Analytical Chemistry (2017-06-13)
Rika Shiota, Hirotoshi Morita, Tomoko Matsumoto, Atsushi Morimoto, Jun Hayakawa, Masako Oka, Hiroshi Kamimori
ANOTACE
An analytical method was developed and validated for the measurement of hydroxyproline (Hyp) levels in mouse kidney by high-performance liquid chromatography with tandem mass spectrometric detection (LC/MS/MS) using an analytical column specially designed for the LC/MS/MS analysis for intact amino acids. Tissue hydrolyzed with hydrochloric acid could be directly injected into the LC/MS/MS, as well as separated and detected using the deuterium labelled Hyp as an internal standard. The calibration curve showed good linearity from 5 to 500 nmol/mg of tissue; the precision and accuracy, including within- and between-run, were less than 6% and within 100 ± 6%, respectively.
MATERIÁLY
Číslo produktu
Značka
Popis produktu
Sigma-Aldrich
L-Proline-2,5,5-d3, 98 atom % D, 97%