Přejít k obsahu
Merck
  • Development of Safe and Potent Oil-in-Water Emulsion of Paclitaxel to Treat Peritoneal Dissemination.
Development of Safe and Potent Oil-in-Water Emulsion of Paclitaxel to Treat Peritoneal Dissemination.
Journal of pharmaceutical sciences (2017-01-09)
Ken-Ichi Ogawara, Yoshiko Fukuoka, Yuta Yoshizawa, Toshikiro Kimura, Kazutaka Higaki
ANOTACE
To develop a safer and more potent paclitaxel (PTX) formulation, we prepared various oil-in-water emulsions by using egg phosphatidylcholine, Tween 80, and a mixture of triglycerides with different fatty acid chain lengths as the cosurfactant, surfactant, and oil phase component, respectively. The mean particle diameters of the PTX-emulsions prepared were around 100 nm. The PTX-emulsions did not provoke histamine release from rat mast cells and did not show any significant hemolytic activity, suggesting that PTX-emulsions are biocompatible. In vivo antitumor activity after single intraperitoneal injection of PTX-emulsions to ascites tumor-bearing mice revealed that the formulation containing tricaproin and triacetin (3:1, wt/wt, PTX-emulsion B) significantly prolonged the survival time and suppressed the accumulation of ascites fluid. Two distinct in vitro release studies showed that the release of PTX from emulsion B was significantly faster than those from other preparations. These results indicate that the adequately sustained PTX release would lead to potent in vivo antitumor activity and that PTX-emulsion B would offer an alternative approach to treat peritoneal dissemination.
MATERIÁLY
Číslo produktu
Značka
Popis produktu
Tricaprylin, European Pharmacopoeia (EP) Reference Standard