Přejít k obsahu
Merck
  • Exploring Non-obvious Hydrophobic Binding Pockets on Protein Surfaces: Increasing Affinities in Peptide-Protein Interactions.
Exploring Non-obvious Hydrophobic Binding Pockets on Protein Surfaces: Increasing Affinities in Peptide-Protein Interactions.
Chembiochem : a European journal of chemical biology (2017-04-23)
Aleksandra Balliu, Lars Baltzer
ANOTACE
A 42-residue polypeptide conjugated to a small-molecule organic ligand capable of targeting the phosphorylated side chain of Ser15 was shown to bind glycogen phosphorylase a (GPa) with a K
MATERIÁLY
Číslo produktu
Značka
Popis produktu
Sigma-Aldrich
Fmoc-α-Me-Ala-OH, ≥97.0% (HPLC)