Přejít k obsahu
Merck
  • Enterovirus infection, CXC chemokine ligand 10 (CXCL10), and CXCR3 circuit: a mechanism of accelerated beta-cell failure in fulminant type 1 diabetes.
Enterovirus infection, CXC chemokine ligand 10 (CXCL10), and CXCR3 circuit: a mechanism of accelerated beta-cell failure in fulminant type 1 diabetes.
Diabetes (2009-07-31)
Shoichiro Tanaka, Yoriko Nishida, Kaoru Aida, Taro Maruyama, Akira Shimada, Masako Suzuki, Hiroki Shimura, Soichi Takizawa, Masashi Takahashi, Daiichiro Akiyama, Sayaka Arai-Yamashita, Fumihiko Furuya, Akio Kawaguchi, Masahiro Kaneshige, Ryohei Katoh, Toyoshi Endo, Tetsuro Kobayashi
ANOTACE
Fulminant type 1 diabetes is characterized by the rapid onset of severe hyperglycemia and ketoacidosis, with subsequent poor prognosis of diabetes complications. Causative mechanisms for accelerated beta-cell failure are unclear. Subjects comprised three autopsied patients who died from diabetic ketoacidosis within 2-5 days after onset of fulminant type 1 diabetes. We examined islet cell status, including the presence of enterovirus and chemokine/cytokine/major histocompatibility complex (MHC) expressions in the pancreata using immunohistochemical analyses and RT-PCR. Immunohistochemical analysis revealed the presence of enterovirus-capsid protein in all three affected pancreata. Extensive infiltration of CXCR3 receptor-bearing T-cells and macrophages into islets was observed. Dendritic cells were stained in and around the islets. Specifically, interferon-gamma and CXC chemokine ligand 10 (CXCL10) were strongly coexpressed in all subtypes of islet cells, including beta-cells and alpha-cells. No CXCL10 was expressed in exocrine pancreas. Serum levels of CXCL10 were increased. Expression of MHC class II and hyperexpression of MHC class I was observed in some islet cells. These results strongly suggest the presence of a circuit for the destruction of beta-cells in fulminant type 1 diabetes. Enterovirus infection of the pancreas initiates coexpression of interferon-gamma and CXCL10 in beta-cells. CXCL10 secreted from beta-cells activates and attracts autoreactive T-cells and macrophages to the islets via CXCR3. These infiltrating autoreactive T-cells and macrophages release inflammatory cytokines including interferon-gamma in the islets, not only damaging beta-cells but also accelerating CXCL10 generation in residual beta-cells and thus further activating cell-mediated autoimmunity until all beta-cells have been destroyed.