- Immune-Induced Fever Is Dependent on Local But Not Generalized Prostaglandin E
Immune-Induced Fever Is Dependent on Local But Not Generalized Prostaglandin E
The Journal of neuroscience : the official journal of the Society for Neuroscience (2017-04-26)
Anna Eskilsson, Takashi Matsuwaki, Kiseko Shionoya, Elahe Mirrasekhian, Joanna Zajdel, Markus Schwaninger, David Engblom, Anders Blomqvist
PMID28438967
ANOTACE
Fever occurs upon binding of prostaglandin E
MATERIÁLY
Číslo produktu
Značka
Popis produktu
Sigma-Aldrich
Dulbecco′s Phosphate Buffered Saline, With MgCl2 and CaCl2, liquid, sterile-filtered, suitable for cell culture